Skip to Content
Merck

Skip To

SCM101

Cardiac Stem Cell Maintenance Medium

The Cardiac Stem Cell Maintenance Medium is available in a 500 mL format & has been optimized & validated for cell culture.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 mL

Estimated to ship onJune 09, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 6,730.00

About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
eCl@ss:
32160801

CZK 6,730.00


Estimated to ship onJune 09, 2026Details


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

technique(s)

cell culture | mammalian: suitable

input

sample type cardiac stem cell(s)

shipped in

dry ice

storage temp.

-10 to -25°C

General description

The Cardiac Stem Cell Maintenance Medium has been optimized and qualified for the growth and expansion of cardiac stem cells derived from rodent origins. Cells expanded in Cardiac Stem Cell Maintenance Medium express the correct cardiac stem cell markers and are furthermore capable of differentiating into cardiomyocytes.

Physical form

Cardiac Stem Cell Maintenance Medium is a proprietary formulation that contains fetal bovine serum.

Preparation Note

For long term storage, maintain the media at -20°C. Prior to initial use, thaw frozen media at 4°C overnight or until it has become completely equilibrated. Maintain thawed media at 2-8°C in the dark for up to one month.

Analysis Note

Each lot of medium is tested for the optimal cell growth and proliferation of cardiac stem cells. Sterility Testing: Negative

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SCM102SCM003SCM005
form

liquid

form

liquid

form

liquid

form

liquid

technique(s)

cell culture | mammalian: suitable

technique(s)

cell differentiation: suitable

technique(s)

cell culture | stem cell: suitable

technique(s)

cell culture | stem cell: suitable

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

storage temp.

-10 to -25°C

storage temp.

-10 to -25°C

storage temp.

-10 to -25°C

storage temp.

-10 to -25°C

Quality Level

100

Quality Level

100

Quality Level

300

Quality Level

100

input

sample type cardiac stem cell(s)

input

sample type cardiac stem cell(s)

input

sample type neural stem cell(s)

input

sample type neural stem cell(s)


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters