Skip to Content
Merck

Skip To

SCM003

Neural Stem Cell Basal Medium

The Neural Stem Cell Basal Medium is a defined serum-free, growth factor-free medium that has been optimized for the growth & in vitro differentiation of neural stem cells derived from rodents.

Synonym(s):

NSC Culture Media

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 mL

Available to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

CZK 5,930.00

About This Item

UNSPSC Code:
12352207
NACRES:
NA.71
eCl@ss:
32160801

CZK 5,930.00


Available to ship TODAYDetails


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

manufacturer/tradename

Chemicon®

technique(s)

cell culture | stem cell: suitable

input

sample type neural stem cell(s)

shipped in

dry ice

storage temp.

-10 to -25°C

General description

The Neural Stem Cell Basal Medium is a defined serum-free, growth factor-free medium that has been optimized for the growth and in vitro differentiation of neural stem cells derived from rodents. When used in conjunction with bFGF or bFGF, EGF and heparin, the basal medium will allow for the proliferation of rat and mouse neural stem cells, respectively. Withdrawal of the growth factors from the Neural Stem Cell Basal Medium will result in the spontaneous differentiation of rodent neural stem cells.

Application

The Neural Stem Cell Basal Medium is a defined serum-free, growth factor-free medium that has been optimized for the growth & in vitro differentiation of neural stem cells derived from rodents.

Preparation Note

Store at -20°C until ready to use. Upon thawing, the basal medium should be stored at 2-8°C for up to one month.

Analysis Note

Each lot of medium is tested for the optimal cell growth, proliferation and differentiation of both rat and mouse neural stem cells. Sterility Testing: Negative; Osmolarity: 299; pH: 7.4

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SCM005SCM004SCM007
technique(s)

cell culture | stem cell: suitable

technique(s)

cell culture | stem cell: suitable

technique(s)

cell culture | stem cell: suitable

technique(s)

cell culture | stem cell: suitable

form

liquid

form

liquid

form

liquid

form

liquid

storage temp.

-10 to -25°C

storage temp.

-10 to -25°C

storage temp.

-10 to -25°C

storage temp.

-10 to -25°C

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

Quality Level

300

Quality Level

100

Quality Level

100

Quality Level

100

manufacturer/tradename

Chemicon®

manufacturer/tradename

Chemicon®

manufacturer/tradename

Chemicon®

manufacturer/tradename

Chemicon®


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. Could you please provide the detailed composition/component of neural stem cell basal medium.

    1 answer
    1. The specific formulation of this medium is considered proprietary. The general composition is as follows:
      DMEM/F12 without HEPES
      L-Glutamine
      Penicillin/Streptomycin
      B27 Supplement, 1X

      Please see the link below to review additional information available in the product datasheet:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/227/239/scm003.pdf

      Helpful?

Reviews

No rating value

Active Filters