Skip to Content
Merck

Skip To

SCC289

SF295 Human Glioblastoma Cell Line

SF295 human glioblastoma cell line is a valuable model for glioblastoma research.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 vial
Please contact Customer Service for Availability
CZK 30,400.00

About This Item

NACRES:
NA.45
UNSPSC Code:
41106514

CZK 30,400.00


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


application(s)

cell analysis

General description

Glioblastoma multiforme is the most common and malignant primary brain tumor in adults, with a high recurrence rate and a five-year survival rate of less than 5% (1). Nearly 50% of glioblastomas originate in the frontal or temporal lobes of the brain (1). Glioblastomas are the most aggressive form of cancer and demonstrate a high degree of chemotherapy resistance, highlighting the importance of relevant human patient-derived cell models for advancing research into characteristics and treatment of this disease.

SF295 is a patient-derived glioblastoma cell line originating from a temporal lobe tumor (2). SF295 cells do not express the glial markers GFAP or glutamine synthetase, but have been shown to positively express neurofibromin-1, identified as one of the most frequently mutated factors in glioblastoma multiforme (3). SF295 cells exhibit hypertriploidy and glial-like morphology in culture and are non-tumorigenic in athymic mice (2). The SF295 cell line is highly published in studies of chemotherapeutic treatments for glioblastoma, representing a valuable cellular model for assessment of drugs and molecular modulators for their effect on glioblastomas.

<bold>Source:</bold>
The SF295 glioblastoma cell line was derived from the left frontal lobe tumor of a 67-year-old female patient (2).

Research Category:
Cancer

Biochem/physiol Actions

Human, Cancer Cells

Packaging

&ge;1X106 cells/vial

Preparation Note

Store in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Other Notes

This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact [email protected].

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SCC282SCC173SCC283
application(s)

cell analysis

application(s)

cell analysis

application(s)

-

application(s)

cell analysis


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

212.0 °F

flash_point_c

100 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
SCC28904061842104031

Questions

Reviews

No rating value

Active Filters