Skip to Content
Merck

Skip To

PP63

IgG, Rabbit Gamma Globulin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 kit

Estimated to ship onApril 21, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 19,200.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.71
eCl@ss:
32160702
Clone:
polyclonal
Species reactivity:
rabbit
Application:
Citations:
-

CZK 19,200.00


Estimated to ship onApril 21, 2026Details


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


antibody form

purified antibody

Quality Level

clone

polyclonal

species reactivity

rabbit

Preparation Note

Maintain frozen at -20°C in undiluted aliquots for up to 6months.

Disclaimer

This product is intended for use by IVD manufacturers.
Contact customer service for assistance: [email protected]
Minimum order quantity required to qualify. Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
PP63-K04053252266782
PP63-10-KC04053252523786