Skip to Content
Merck
All Photos(1)

Key Documents

MABF935

Sigma-Aldrich

Anti-MPR300/IGF-2R/CD222 Antibody, clone 2C2

clone 2C2, from mouse

Synonym(s):

Cation-independent mannose-6-phosphate receptor, 300 kDa mannose 6-phosphate receptor, CD222, CI Man-6-P receptor, CI-MPR, IGF-II receptor, Insulin-like growth factor 2 receptor, Insulin-like growth factor II receptor, M6P/IGF2 receptor, M6P/IGF2R, M6P-R

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CZK 10,300.00

CZK 10,300.00


Estimated to ship onApril 14, 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
CZK 10,300.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CZK 10,300.00


Estimated to ship onApril 14, 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2C2, monoclonal

species reactivity

human

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... IGF2R(3482)

General description

Cation-independent mannose-6-phosphate receptor (UniProt P11717; also known as 300 kDa mannose 6-phosphate receptor, CD222, CI Man-6-P receptor, CI-MPR, IGF-II receptor, Insulin-like growth factor 2 receptor, Insulin-like growth factor II receptor, M6P/IGF2 receptor, M6P/IGF2R, M6P-R, M6PR, MPR 300) is encoded by the IGF2R (also known as CIMPR, MPRI) gene (Gene ID 3482) in human. Mannose 6-phosphate (M6P) receptors (MPRs) mediate the delivery of newly synthesized, mannose 6-phosphate-bound lysosomal hydrolases from the secretory pathway to the endosomal system. MPRs bind lysosomal hydrolases in the trans-Golgi network (TGN) and unload the enzyme in the endosomal system. There exist a cation-independent CI-MPR (IGF2R) and a smaller CD-MPR that requires divalent cations for lysosomal hydrolases recognition. MPRs also undergo recycling between the endosomal system and the plasma membrane, where CI-MPR mediates the internalization of other ligands, including insulin-like growth factor II (IGF-II). CI-MPR/IGF2R is produced with a signal peptide (a.a. 1-40), the removal of which yields the mature receptor with a large lumenal/extracellular region (a.a. 41-2304), a transmembrane segment (a.a. 2305-2327), and a cytoplasmic domain (a.a. 2328-2491), The extracellular region is composed of 15 P-type carbohydrate recognition domains, also referred to as MRH (mannose 6-phosphate receptor homology) domains. Domains 3 and 9 bind M6P with high affinity and domain 5 binds M6P with weak affinity, while domain 11 binds IGF-II.

Specificity

Clone 2C2 targets the extracellular (lumenal) domain of IGF-II receptor (MPR). Clone 2C2 competed against IGF-II for MPR binding without affecting MPR interaction with pentamannose 6-phosphate (PMP) or beta-hexosaminidase, nor MPR-mediated beta-hexosaminidase endocytosis (Braulke, T., et al. (1988). Biochem. Res. Commun. 150(3):1287-1293; Braulke, T., et al. (1987). J. Cell Biol.;104(6):1735-1742).

Immunogen

Epitope: Extracellular (lumenal) domain.
Recombinant full-length human MPR300/IGF-2R/CD222.

Application

Affinity Binding Assay: Representative lots were labeled with 125I and employed to bind and assess IGF-II receptor (MPR) on the surface of human fibroblasts (van Rahden, V.A., et al. (2012). Hum. Mol. Genet. 21(23):5019-5038; Braulke, T., et al. (1987). J. Cell Biol.;104(6):1735-1742).
Affinity Binding Assay: A representative lot competed against IGF-II, but not pentamannose 6-phosphate (PMP), for binding immobilized IGF-II receptor (MPR) (Braulke, T., et al. (1988). Biochem. Res. Commun. 150(3):1287-1293).
Western Blotting Analysis: A representative lot was labeled with 125I and detected the ~300 kDa IGF-II receptor (MPR300) in lysates from HeLa cells and primary human macrophages (Pohl, S., et al. (2010). J. Biol. Chem. 285(31):23936-23944).
Immunohistochemistry Analysis: A representative lot detected hepatocyte membrane and cytoplasmic M6P/IGF-2R immunoreactivity in normal human liver frozen sections. A reduced immunostaining of hepatocytes restricted to the sinusoidal part of the cell membrane and an increased signals in perisinusoidal cells was seen in cirrhotic livers (Sedlaczek, N., et al. (2003). Br. J. Cancer. 88(5):733-739).
Immunocytochemistry Analysis: A representative lot immunostained late endosomes in 2% paraformaldehyde-fixed, 0.01% saponin-permeabilized human fibroblasts (Bakker, A.C., et al. (1997). J. Cell Sci. 110 (Pt 18):2227-2238).
Anti-MPR300/IGF-2R/CD222 Antibody, clone 2C2 is an antibody against MPR300/IGF-2R/CD222 for use in Immunocytochemistry, Immunohistochemistry, Affinity Binding Assay, Western Blotting.
Research Category
Inflammation & Immunology
Research Sub Category
Vesicular Trafficking

Quality

Evaluated by Immunocytochemistry in HeLa cells.

Immunocytochemistry Analysis: A 1:250 dilution of this antibody detected MPR300/IGF-2R/CD222 in HeLa cells.

Target description

270.3 calculated. ~300 kDa reported (Pohl, S., et al. (2010). J. Biol. Chem. 285(31):23936-23944).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service