Skip to Content
Merck
All Photos(2)

Key Documents

MABT1499

Sigma-Aldrich

Anti-MBP (Citrulline R122) Antibody, clone 1H1

clone 1H1, from mouse

Synonym(s):

Myelin basic protein, Myelin A1 protein, Myelin membrane encephalitogenic protein, Citrullinated MBP

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
CZK 3,890.00
100 μG
CZK 10,700.00

CZK 3,890.00


Estimated to ship onApril 01, 2025Details


Request a Bulk Order

Select a Size

Change View
25 μG
CZK 3,890.00
100 μG
CZK 10,700.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CZK 3,890.00


Estimated to ship onApril 01, 2025Details


Request a Bulk Order

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1H1, monoclonal

species reactivity

mouse, human

packaging

antibody small pack of 25 μg

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

citrullination (Arg122)

General description

Myelin basic protein (UniProt: P02686; also known as MBP, Myelin A1 protein, Myelin membrane encephalitogenic protein) is encoded by the MBP gene (Gene ID: 4155) in human. MBP is a homodimeric protein that is found in both the central and the peripheral nervous system. It is one of the most abundant protein component of the myelin membrane in the CNS and plays a role in both the formation and stabilization of this myelin membrane. It is responsible for adhesion of the cytosolic surfaces of multilayered compact myelin. MBP can interact with many polyanionic proteins including actin, tubulin, calmodulin, and clathrin, and with negatively charged lipids. MBP isoforms are found in both the central and the peripheral nervous system, whereas Golli-MBP isoforms are expressed in fetal thymus, spleen and spinal cord, as well as in cell lines derived from the immune system. MBP expression begins abruptly in 14-16-week-old fetuses. Even smaller isoforms seem to be produced during embryogenesis; some of these persisting in the adult. MBP citrullination by peptidylarginine deiminase (PAD) leads to incomplete protein-lipid bilayer interactions and increases its vulnerability to proteolytic enzymes, resulting in disorganization of the myelin sheath in the central nervous system. Citrullinated MBP (citMBP) has been suggested to be a hallmark of demyelination. MBP citrullination has frequently been demonstrated in various CNS disorders. citMBP at R25, R122, and R130 is reported in the brains of subjects with Creutzfeldt-Jakob disease (CJD). Clone 1H1 specifically recognizes citMBP at R122. (Ref.: Jang, B., et al. (2017). Mol. Neurobiol. 55(4); 3172-3184; Boggs, JM (2006). Cell. Mol. Life Sci. 63(17); 1945-1961).

Specificity

Clone 1H1 is a mouse monoclonal antibody that detects human and murine myelin basic protein citrullinated on residue 122. It targets an epitope with in 19 amino acids from the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 19 amino acids from isoform 5 of human Myelin Basic Protein with citrullination on residue 122.

Application

Anti-MBP (Citrulline R122), clone 1H1, Cat. No. MABT1499, is a mouse monoclonal antibody that detects Myelin basic protein citrullinated on residue 122 and has been is tested for use in ELISA, Immunohistochemistry, and Western Blotting.
ELISA Analysis: A representative lot detected MBP (Citrulline R122) in ELISA applications (Jang, B., et. al. (2018). Mol Neurobiol. 55(4):3172-3184).

Western Blotting Analysis: A representative lot detected MBP (Citrulline R122) in Western Blotting applications (Jang, B., et. al. (2018). Mol Neurobiol. 55(4):3172-3184).

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected MBP (Citrulline R122) in Myelin basic protein (MBP) treated with human PAD2 (Courtesy of Byungki Jang, Ph.D./Eun-Kyoung Choi, Ph.D., Ilsong Institute of Life Science, Hallym University, South Korea).





Immunohistochemistry Analysis: A representative lot detected MBP (Citrulline R122) in Immunohistochemistry applications (Jang, B., et. al. (2018). Mol Neurobiol. 55(4):3172-3184).

(Courtesy of Byungki Jang, Ph.D./Eun-Kyoung Choi, Ph.D., Industry Academic Cooperation Foundation, Hallym University, South Korea).
Research Category
Cell Structure

Quality

Evaluated by Western Blotting in GST-tagged myelin basic protein (MBP) treated with human PAD-2.

Western Blotting Analysis: 1 µg/mL of this antibody detected MBP (Citrulline R122) in GST-tagged myelin basic protein (MBP) treated with human Peptidyl Arginine Deiminase 2 (PAD-2)(200 ng for 4 h at 37°C).

Target description

~41 kDa observed; 18.59 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tiemei Li et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 10(22), e2206432-e2206432 (2023-05-29)
Pneumonia is one of the leading causes of death in patients with acute ischemic stroke (AIS). Antibiotics fail to improve prognosis of patients with post-stroke pneumonia, albeit suppressing infection, due to adverse impacts on the immune system. The current study

Questions

  1. Could you provide the tris-glycine buffer concentration for MABT1499?

    1 answer
    1. The formulation is similar to a 1X solution of Tris-Glycine buffer solutions. If you need more information, you can visit the Sigma-Aldrich website and search for "TRIS Glycine buffer solution BioUltra, 10× concentrate" and "Tris-Glycine Buffer 10× Concentrate."

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service