Skip to Content
Merck

Skip To

MABF604

Anti-CD45R (B220) Antibody (human/mouse), PE-Cy7, clone RA3-6B2

clone RA3-6B2, 0.2 mg/mL, from rat

Synonym(s):

Receptor-type tyrosine-protein phosphatase C, Leukocyte common antigen, L-CA, Lymphocyte antigen 5, Ly-5, T200, CD45

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
PE-Cy7
Clone:
RA3-6B2, monoclonal
Application:
FACS, IF, IHC, IP, WB
Citations:
-
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rat

Quality Level

conjugate

PE-Cy7

antibody form

purified antibody

antibody product type

primary antibodies

clone

RA3-6B2, monoclonal

species reactivity

human, mouse

packaging

antibody small pack of 25 μg

concentration

0.2 mg/mL

technique(s)

flow cytometry: suitable, immunofluorescence: suitable, immunohistochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG2aκ

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PTPRC(5788)
mouse ... Ptprc(19264)

General description

The RA3-6B2 antibody reacts with the human and mouse CD45 isoform known as CD45R, or B220, a protein tyrosine phosphatase of 220 kDa. CD45 is one of the most abundant hematopoietic markers, and is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). Various isoforms are generated and expressed in cell-specific patterns, all critical for leukocyte function. In mouse, the CD45R/B220 isoform is predominantly found on B cells, at varying levels on all stages from pro-B cells to activated B cells, and may also be detected on certain T cell and NK cell subsets. It is of note that B220 is not similarly expressed on human B cells, where it appears to be differentiation-specific and therefore expressed on only some B cell subsets. Other forms of CD45 with restricted cellular expression include CD45RA, CD45RB, CD45RO and several others.The RA3-6B2 antibody is one of the most consistently used leukocyte markers for B cells, T cell subsets and NK cell subsets in human and mouse. This antibody has also been reported as cross-reactive with feline CD45R/B220.

Immunogen

Immunogen information for this antibody is proprietary information.

Application

Research Category
Inflammation & Immunology
This rat monoclonal antibody Anti-CD45R (B220) (human/mouse), PE-Cy7, clone RA3-6B2 validated for use in western blotting, IP, IHC, Immunofluorescence & Flow Cytometry for the detection of CD45R (B220).

Physical form

Purified Rat Monoclonal Antibody in buffer containing 10 mM NaH2PO4, 150 mM NaCl, 0.1% gelatin, pH 7.2, and up to 0.1% sodium azide.
Size Exclusion Chromatography

Preparation Note

It is recommended to store the product undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.

Analysis Note

The monoclonal antibody was purified from tissue culture supernatant via affinity chromatography. The purified antibody was conjugated under optimal conditions, with unreacted dye removed from the preparation.

Flow Cytometry Analysis: 0.125 µg of this antibody detected CD45R (B220) in one million C57Bl/6 splenocytes.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MABF445MABF444MABF443
clone

RA3-6B2, monoclonal

clone

RA3-6B2, monoclonal

clone

RA3-6B2, monoclonal

clone

RA3-6B2, monoclonal

species reactivity

human, mouse

species reactivity

human, mouse

species reactivity

mouse, human

species reactivity

human, mouse

Gene Information

human ... PTPRC(5788)
mouse ... Ptprc(19264)

Gene Information

human ... PTPRC(5788)
mouse ... Ptprc(19264)

Gene Information

human ... PTPRC(5788)
mouse ... Ptprc(19264)

Gene Information

human ... PTPRC(5788)
mouse ... Ptprc(19264)

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

conjugate

PE-Cy7

conjugate

PE-Cy7

conjugate

PE-Cy5

conjugate

PE

biological source

rat

biological source

rat

biological source

rat

biological source

rat


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
MABF60404053252965708

Questions

Reviews

No rating value

Active Filters