Skip to Content
Merck
All Photos(3)

Key Documents

MABF3180

Sigma-Aldrich

Anti-CCL4/MIP1β Antibody, clone 4G11.10.5

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
CZK 3,910.00
100 μG
CZK 9,860.00

CZK 3,910.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
25 μG
CZK 3,910.00
100 μG
CZK 9,860.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.74

CZK 3,910.00


In StockDetails


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

4G11.10.5, monoclonal

mol wt

calculated mol wt 10.21 kDa
observed mol wt ~8.0 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG2aκ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... CCL4(6351)

General description

C-C motif chemokine 4 (UniProt: P13236; also known as G-26 T-lymphocyte-secreted protein, HC21, Lymphocyte activation gene 1 protein, LAG-1, MIP-1-beta (1-69),Macrophage inflammatory protein 1-beta, MIP-1-beta, PAT 744, Protein H400, SIS-gamma, Small-inducible cytokine A4, T-cell activation protein 2, ACT-2) is encoded by the CCL (also known as LAG1, MIP1B, SCYA4) gene (Gene ID: 6351) in human. CCL4/MIP1b is a secretory protein that is synthesized with a signal peptide (aa 1-23), which is subsequently cleaved off to produce the mature form (aa 24-292) that can be further proteolytically cleaved at the N-terminal to generate MIP-1b (aa 26-92). It is present either as a homodimer or a heterodimer of MPIa and MIP1b. Both MIP-1a and MIP1β participate in the host response to invading bacterial, viral, parasite and fungal pathogens by regulating the trafficking and activation state of selected subgroups of inflammatory cells. Although they exert similar effects on monocytes their effect on lymphocytes differs; with MIP1a selectively attracting CD8+ lymphocytes and MIP1β selectively attracting CD4+ lymphocytes. MIP1b serves as a monokine with inflammatory and chemokinetic properties. It is one of the major HIV-suppressive factors produced by CD8+ T-cells that inhibits different strains of HIV-1, HIV-2, and simian immunodeficiency virus. CCR5 serves as a specific receptor for MIP1b. The processed form MIP-1b also retains the ability to induce down-modulation of surface expression of the chemokine receptor CCR5 and inhibit the CCR5-mediated entry of HIV-1 in T-cells. It also serves as a ligand for CCR1 and CCR2 isoform B.

Specificity

Clone 4G11.10.5 is a mouse monoclonal antibody that detects human C-C motif chemokine 4 (CCL4/MIP1b ).

Immunogen

Full-length human recombinant C-C-motif chemokine 4 (CCL4) (without the signal peptide).

Application

Quality Control Testing

Evaluated by Western Blotting with human recombinant C-C motif chemokine 4 (CCL4).

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected recombinant human CCL4/MIP1b .

Tested Applications

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution from a representative lot detected CCL4/MIP1b a in human small intestine and human adrenal gland tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-CCL4/MIP1β, clone 4G11.10.5, Cat. No. MABF3180, is a mouse monoclonal antibody that detects CCL4/ MIP1β and is tested for use in Immunohistochemistry (Paraffin) and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG2a in PBS with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service