Skip to Content
Merck
All Photos(1)

Key Documents

MABF1977

Sigma-Aldrich

Anti-phospho-C5aR/CD88 (Ser334) Antibody, clone 32-G1

clone 32-G1, from mouse

Synonym(s):

C5a anaphylatoxin chemotactic receptor 1, C5a anaphylatoxin chemotactic receptor, C5a-R, C5aR, CD88

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CZK 9,510.00

CZK 9,510.00


Estimated to ship onMay 14, 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
CZK 9,510.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CZK 9,510.00


Estimated to ship onMay 14, 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

32-G1, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

phosphorylation (pSer334)

Gene Information

human ... C5AR1(728)

General description

C5a anaphylatoxin chemotactic receptor 1 (UniProt: P21730; also known as C5a anaphylatoxin chemotactic receptor, C5a-R, C5aR, CD88) is encoded by the C5AR1 (also known as C5AR, C5R1) gene (Gene ID: 728) in human. C5aR is a multi-pass membrane protein that serves as a receptor for the chemotactic and inflammatory peptide anaphylatoxin C5a. It exists as a homodimer, but can also form higher-order oligomers. C5a interacts with at least two sites on the receptor: a high-affinity site on the extracellular N-terminus, and a second site in the transmembrane region, which activates downstream signaling events. Receptor activation stimulates chemotaxis, granule enzyme release, and intracellular calcium release and superoxide anion production. Chemotaxis inhibitory protein of Staphylococcus aureus (CHIPS), a 121-residue protein, is reported to bind to the N-terminus of the C5aR (aa 1-35) with nanomolar affinity and inhibits C5a-mediated responses in human leukocytes. Sulfation at Tyrosine 14 is shown to be important for CHIPS binding. Upon C5a binding, C5aR) undergoes rapid phosphorylation on the six serine residues present in the C-terminal region followed by desensitization and internalization. The key residues involved in this process are Serine 334 and Serine 338. Phosphorylated C5aR is shown to co-localize with beta-arrestin 1 and 2 in cytoplasmic vesicles. Although C5aR activation is able to promote a loose association with beta-arrestins, but phosphorylation of either Serine 334)/338 or Serine 327)/338 is reported to be essential for the formation of a persistent complex. (Ref.: Ippel, JH., et al., (2009). J. Biol. Chem. 284(18); 12363-12372; Braun, L., et al. (2003). J. Biol. Chem. 278(6); 4277-4285).

Specificity

Clone 32-G1 specifically detects C5a anaphylatoxin chemotactic receptor 1 (C5aR) phosphorylated on Ser334. It targets an epitope within 18 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 18 amino acids surrounding phophorylated Serines 332, 334, and 338 from the C-terminal region of human C5a anaphylatoxin chemotactic receptor 1 (C5aR).

Application

Anti-phospho-C5aR_CD88 (Ser334), clone 32-G1, Cat. No. MABF1977, is a mouse monoclonal antibody that detects C5a anaphylatoxin chemotactic receptor 1 (C5aR1) and has been tested for use in ELISA, Immunofluorescence, and Western Blotting.
Research Category
Inflammation & Immunology
Western Blotting Analysis: A representative lot detected phospho-C5aR_CD88 (Ser334) in Western Blotting applications (Pollok-Kopp, B., et. al. (2007). J Biol Chem. 282(7):4345-53).

Enzyme Immunoassay Analysis: A representative lot detected phospho-C5aR_CD88 (Ser334) in Enzyme Immunoassay applications (Pollok-Kopp, B., et. al. (2007). J Biol Chem. 282(7):4345-53).

Quality

Evaluated by Western Blotting using a mixture of two C5aR peptides corresponding to amino acids 327-344 (phosphorylated on Serine 332, 334, and 338) and amino acids 1-31 conjugated to BSA vs. irrelevant peptides conjugated to BSA (negative control).

Western Blotting Analysis: 1:20,000 of this antibody detected phospho-C5aR_CD88 (Ser334) in a mixture of two C5aR peptides corresponding to amino acids 327-344 (phosphorylated at Serine 332, 334, and 338) and amino acids 1-31, conjugated to BSA vs. irrelevant peptides conjugated to BSA (negative control).

Target description

~100-110 kDa observed; 39.34 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service