Skip to Content
Merck
All Photos(1)

Documents

MABD150

Sigma-Aldrich

Anti-LIF Receptor, clone 1C7 (Azide Free) Antibody

clone 1C7, from mouse

Synonym(s):

Leukemia inhibitory factor receptor, LIF receptor, LIF-R, D-factor, CD118, GP190, LIFR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1C7, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable
inhibition assay: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LIFR(3977)

General description

The LIF receptor is critical in modern mouse stem cell maintenance as the receptor for LIF, the embryonic stem cell (ESC) growth factor that maintains pluripotency and self renewal in mouse ES cells, however physiologically, the importance of LIF-R in real mouse development has been less clear. LIF receptor does play a signaling role and when cleaved its extracellular portion can act as a decoy receptor and block exogenous LIF from performing its function on ES cells. LIF receptor is expressed in multiple tissues in the mouse during development from the liver to the lung and brain. LIF receptor also is important in immune responses and in the stimulation of the interferon response to viral infection. Also LIF receptor has recently been identified as a metastasis suppressor that functions through the Hippo-YAP pathway in humans.

Immunogen

Recombinant protein corresponding to mouse LIF Receptor.

Application

Anti-LIF Receptor, clone 1C7 (Azide Free) is an antibody against LIF Receptor for use in FC, Inhibition.
Inhibition Assay Analysis: A representative lot detected LIF Receptor in LIF binding to JAR cells (Blanchard, F., et al. (1997) International Immunology. 9(12):1775-1784).
Inhibition Assay Analysis: A representative lot detected LIF Receptor in IL3 proliferative response in Ba/F3 cells (Pitard, V., et al. (1997) Journal of Immunological Methods. 205:177-190).
Research Category
Stem Cell Research
Research Sub Category
Cell Cycle, DNA Replication & Repair

Quality

Evaluated by Flow Cytometry in CHO D1IgD2-mycDAF cells .

Flow Cytometry Analysis: 1.0 µg of this antibody detected LIF Receptor in CHO D1IgD2-mycDAF.

Target description

123 kDa calculated

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nazanin Vaziri et al.
Life (Basel, Switzerland), 11(12) (2021-12-25)
Leukemia inhibitory factor (LIF), as a member of the interleukin-6 cytokine family, plays a complex role in solid tumors. However, the effect of LIF as a tumor microenvironment factor on plasticity control in breast cancer remains largely unknown. In this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service