Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 100 μg | Estimated to ship onMay 15, 2026fromAreál Kühne+Nagel spol. s r.o. | CZK 11,500.00 |
About This Item
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
2G7.1, monoclonal
species reactivity
human
technique(s)
immunohistochemistry: suitable, western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... HELLS(3070)
General description
Immunogen
Application
Epigenetics & Nuclear Function
Signaling Neuroscience
Physical form
Preparation Note
Analysis Note
Western Blotting Analysis: 1.0 µg/mL of this antibody detected Lymphoid-specific helicase (SMARCA6) in 10 µg of human thymus tissue lysate.
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| biological source mouse | biological source rat | biological source mouse | biological source mouse |
| Gene Information human ... HELLS(3070) | Gene Information human ... SMARCA1(6594) | Gene Information human ... SMARCE1(6605) | Gene Information human ... SMARCD2(6603) |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| species reactivity human | species reactivity human | species reactivity human | species reactivity human |
| clone 2G7.1, monoclonal | clone SNF 2C4, monoclonal | clone 6G11, monoclonal | clone 2F7, monoclonal |
| antibody form purified antibody | antibody form purified immunoglobulin | antibody form purified immunoglobulin | antibody form purified immunoglobulin |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| MABE421 | 04053252966088 |



