Skip to Content
Merck

Skip To

MABE330

Anti-c-ErbB2/c-Neu Antibody, clone 3B5

clone 3B5, from mouse

Synonym(s):

Receptor tyrosine-protein kinase erbB-2, Metastatic lymph node gene 19 protein, MLN 19, Proto-oncogene Neu, Proto-oncogene c-ErbB-2, Tyrosine kinase-type cell surface receptor HER2, p185erbB2, CD340

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μg
Check Cart for Availability
CZK 11,800.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
3B5, monoclonal
Application:
IHC, WB
Citations:
-

CZK 11,800.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1245

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3B5, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable, western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ERBB2(2064)

General description

The ErbB2 (HER2, c-Neu, ErbB2, c-erbB2, or HER2/neu) protein is member of the human epidermal growth factor receptor (HER) family. It is a receptor tyrosine kinase containing an intracellular kinase domain and an extracellular ligand domain, although no endogenous ligand has yet been found. ErbB2 dimerizes with ErbB1, 3, and 4, and can activate different pathways including the PI3K, PLCγ, and MAPK pathways, depending on the ErbB receptor involved. Coupling with ErbB3 activates the PI3K pathway, whereas coupling with any of these three receptors can induce the MAPK pathway. ErbB2 signaling promotes cell proliferation and inhibits apoptosis. ErbB2 is overexpressed in various cancers and may play a role in the development and metastasis of gliomas, and ovarian, breast, lung, and gastric cancer.
~185 kDa observed. Uniprot describes four isoforms at ~138 kDa (isoform 1), ~63 kDa (isoform 2), ~71 kDa (isoform 3), and ~137 kDa (isoform 4), which can be highly glycosylated and phosphorylated. This antibody may detect one or more isoforms. Uncharacterized bands may be observed at ~27 kDa and ~38 kDa in some cell lysates.

Immunogen

Linear peptide corresponding human c-ErbB2/c-Neu.

Application

Anti-c-ErbB2/c-Neu Antibody, clone 3B5 is a Mouse Monoclonal Antibody for detection of c-ErbB2/c-Neu also known as MLN 19, Proto-oncogene Neu, Proto-oncogene c-ErbB-2 or CD340, has been validated in WB & IHC.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected c-ErbB2/c-Neu in human breast cancer tissue.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
SK-BR-3 cell lysate
Evaluated by Western Blot in SK-BR-3 cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected c-ErbB2/c-Neu in 10 µg of SK-BR-3 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Replaces: 04-1127

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB1403787SAB5700151OP15
species reactivity

human

species reactivity

human

species reactivity

rat, human, mouse

species reactivity

human, mouse

biological source

mouse

biological source

mouse

biological source

rabbit

biological source

mouse

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

-

clone

3B5, monoclonal

clone

3B1, monoclonal

clone

polyclonal

clone

3B5, monoclonal

antibody form

purified antibody

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

purified antibody

Gene Information

human ... ERBB2(2064)

Gene Information

human ... ERBB2(2064)

Gene Information

human ... ErbB2(2064)

Gene Information

human ... ERBB2(2064)


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

New Products: Antibodies, Assays, Small Molecules, Inhibitors, and Proteins





Global Trade Item Number

SKUGTIN
MABE33004053252522239

Questions

Reviews

No rating value

Active Filters