Skip to Content
Merck

Skip To

MABE1035

Anti-ZBTB46 (ZNF340) Antibody, clone 5A8.1

clone 5A8.1, from mouse

Synonym(s):

Zinc finger and BTB domain-containing protein 46, BTB-ZF protein expressed in effector lymphocytes, BTB/POZ domain-containing protein 4, BZEL, Zinc finger protein 340

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
5A8.1, monoclonal
Application:
IHC, WB
Citations:
-
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5A8.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... ZBTB46(140685)

General description

Zinc finger and BTB domain-containing protein 46 (UniProt Q86UZ6; also known as BTB-ZF protein expressed in effector lymphocytes, BTB/POZ domain-containing protein 4, BZEL, Classical dendritic cell-restricted zinc finger transcription factor zDC, Zinc finger protein 340) is encoded by the ZBTB46 (also known as BTBD4, ZNF340) gene (Gene ID 140685) in human. ZBTB46/ZNF340 is a broad complex, tramtrack, bric-a-brac and zinc finger (BTB-ZF) protein expressed in classical dendritic cells (cDCs), but not other immune cell populations, including plasmacytoid DCs, monocytes, or macrophages. Genome-wide ChIP-seq analysis reveals that ZBTB46/ZNF340 associates with more than 1,000 gene promoters and acts as a transcriptional repressor against many of these target genes. Zbtb46-knockout profoundly alters the cDC subset composition in the spleen in favor of CD8+ DCs, as well as upregulates activation pathways in steady-state cDCs, including elevated creb1 and MHC II expression, and enhances cDC production of vascular endothelial growth factor, leading to increased vascularization of skin-draining lymph nodes. Human ZBTB46/ZNF340 is a 589-a.a. protein containing a BTB domain (a.a. 31-99) and two C2H2-type Zinc fingers (a.a. 418-436, 446-468).
~70 kDa observed. Target band size appears larger than the calculated molecular weight of 64.08 kDa Uncharacterized bands may be observed in some lysate(s).

Immunogen

GST-tagged recombinant internal fragment from the N-terminal half of human ZBTB46 (ZNF340).

Application

Anti-ZBTB46 (ZNF340), clone 5A8.1 Antibody, Cat. No. MABE1035, is a highly specific mouse monoclonal antibody, that targets ZBTB46 and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:50-250 dilution from a representative lot detected ZBTB46 (ZNF340) in human cererbral cortex, kidney, and tonsil tissue sections.

Biochem/physiol Actions

Clone 5A8.1 targets an internal epitope within the N-terminal half of human ZBTB46 (ZNF340).

Physical form

Format: Purified

Analysis Note

Evaluated by Western Blotting in human ovary tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected ZBTB46 (ZNF340) in 10 µg of human ovary tissue lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters