Skip to Content
Merck

Skip To

MABD128

Anti-Cytoplasm Antibody Marker, clone 6F7.2

clone 6F7.1, 0.5 mg/mL, from mouse

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
6F7.1, monoclonal
Species reactivity:
mouse, human
Application:
ICC
Citations:
-
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6F7.1, monoclonal

species reactivity

mouse, human

concentration

0.5 mg/mL

technique(s)

immunocytochemistry: suitable

isotype

IgMκ

shipped in

wet ice

target post-translational modification

unmodified

General description

The cytoplasm is comprised of cytosol, the gel-like substance enclosed within the cell membrane, and cellular organelles, the cell′s internal sub-structures. It is within the cytoplasm that most cellular activities occur including signaling, metabolism, and processes such as cell division. The MABD128 monoclonal antibody has been raised against an enucleated HEK293 cell cytoplasm preparation and effectively identifies both Human and Mouse cytoplasm structures in immunocytochemical analysis. This antibody can serve as general marker for cytoplasm studying overall cell morphology and migration or as a counter stain to use alongside nuclear based antibodies, such as transcription factors.

Immunogen

HEK293 cell cytoplasm.

Application

Anti-Cytoplasm Antibody Marker, clone 6F7.2 is an antibody against Cytoplasm Marker for use in ICC.

Physical form

Format: Purified

Analysis Note

Evaluated by Immunocytochemistry in HeLa, NIH/3T3, HepG2, and A431 cells.

Immunocytochemistry Analysis: A 1:250 dilution of this antibody detected Cytoplasm Marker in HeLa, NIH/3T3, HepG2, and A431 cells.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
MABD12804053252970573

Questions

Reviews

No rating value

Active Filters