Skip to Content
Merck

Skip To

MABC662

Anti-RIPK2 Antibody, clone 1H10.1

clone 1H10.1, from mouse

Synonym(s):

Receptor-interacting serine/threonine-protein kinase 2, CARD-containing interleukin-1 beta-converting enzyme-associated kinase, CARD-containing IL-1 beta ICE-kinase, RIP-like-interacting CLARP kinase, Receptor-interacting protein 2, RIP-2, Tyrosine-prote

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μL
Please contact Customer Service for Availability
CZK 11,200.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
1H10.1, monoclonal
Application:
IHC, WB
Citations:
-

CZK 11,200.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1H10.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable, western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RIPK2(8767)

General description

Receptor-interacting serine/threonine-protein kinase 2 (EC 2.7.11.1; EC 2.7.10.2; UniProt O43353; also known as Receptor-interacting protein 2, CARD-carrying kinase, CARD-containing IL-1 beta ICE-kinase, CARD-containing interleukin-1 beta-converting enzyme-associated kinase, Growth-inhibiting gene 30, Receptor-interacting protein-like interacting caspase-like apoptosis regulatory protein kinase, RIP-2, RIP-like interacting CLARP kinase, Tyrosine-protein kinase RIPK2) is encoded by the RIPK2 (also known as CARD3, CARDIAK, CCK, GIG30, RICK, RIP2) gene (Gene ID 8767) in human. RIPK2 is a dual-specificity kinase that exhibits both serine/threonine as well as tyrosine kinase activities. RIPK2 participates in transducing cellular inflammatory response by propagating signaling downstream of the intracellular peptidoglycan sensors NOD1 and NOD2.
~60 kDa observed

Immunogen

GST-tagged recombinant protein corresponding to human RIPK2.

Application

Anti-RIPK2 Antibody, clone 1H10.1 is an antibody against RIPK2 for use in Western Blotting and Immunohistochemistry.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected RIPK2 in human prostate and human placenta tissue.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Biochem/physiol Actions

Expected to react with both isoforms.

Physical form

Format: Purified
Purified mouse monoclonal IgMκ antibody in PBS with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in human placenta tissue lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected RIPK2 in 10 µg of human placenta tissue lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB4503468HPA015273MABC82
species reactivity

human

species reactivity

human, mouse

species reactivity

human

species reactivity

human

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Gene Information

human ... RIPK2(8767)

Gene Information

human ... RIPK2(8767)

Gene Information

human ... RIPK2(8767)

Gene Information

human ... RIPK1(8737)

biological source

mouse

biological source

rabbit

biological source

rabbit

biological source

mouse

clone

1H10.1, monoclonal

clone

polyclonal

clone

polyclonal

clone

3C6.1, monoclonal


Still not finding the right product?

Try our Product Selector Tool to narrow your options


wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

12 - Non Combustible Liquids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

A major focus of breast cancer research is to understand the mechanisms responsible for disease progression and drug resistance. Toward that end, it has been found that approximately two thirds of all human breast carcinomas overexpress the Estrogen Receptor α (ERα) protein and it remains the primary pharmacological target for endocrine therapy1,2. The normal cellular function of ERα is as a transcription factor that mediates a wide variety of physiological processes, many of which are dependent upon phosphorylation of the receptor at specific amino acid residues3,4. Indeed, ERα is known to be phosphorylated at a multitude of different sites, yet how these all correlate to disease remains unclear5. Here, we interrogated multiple sites of ERα for phosphorylation status by screening an extensive panel of different breast cancer patient samples and other non-breast cancer tissue microarray (TMA) slide samples to determine their relevance to disease.






Global Trade Item Number

SKUGTIN
MABC66204055977170672

Questions

Reviews

No rating value

Active Filters