Skip to Content
Merck
All Photos(1)

Documents

MAB8262F-5

Sigma-Aldrich

Anti-RSV Antibody, fusion protein, all type A, B strains, clone 133-1H

clone 133-1H, Chemicon®, from mouse

Synonym(s):

RSV

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.45

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified antibody

antibody product type

primary antibodies

clone

133-1H, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

isotype

IgG2aκ

shipped in

wet ice

Specificity

Directed against the 47-49 kDa Fusion Protein of RSV. Antibody MAB858-1 recognizes both A and B RSV strains.

Immunogen

Respiratory Syncytial virus A2

Application

Anti-RSV Antibody, fusion protein, all type A, B strains, clone 133-1H detects level of Respiratory Syncytial Virus & has been published & validated for use in IF.
ELISA, Indirect Immunofluorescence

Final working dilutions must be determined by end user.

Optimal working dilutions must be determined by the end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Purified. Liquid 0.02MPB, 0.25M NaCl, 0.1% sodium azide

Storage and Stability

Store at 2-8ºC

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service