Skip to Content
Merck

Skip To

MAB4306

Anti-Human Leukemia Inhibitory Factor Antibody, clone 4F7.2

clone 4F7.2, Chemicon®, from mouse

Synonym(s):

hLIF

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg

Estimated to ship onJune 03, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 11,600.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
4F7.2, monoclonal
Application:
ELISA, WB
Citations:
3

CZK 11,600.00


Estimated to ship onJune 03, 2026Details


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4F7.2, monoclonal

species reactivity

mouse, human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable, western blot: suitable

input

sample type neural stem cell(s)
sample type hematopoietic stem cell(s)

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LIF(3976)

General description

Leukemia inhibitory factor, or LIF, is a pleiotropic cytokine that is expressed by a wide variety of cells including activated T lymphocytes, monocytes, mast cells and neuronal cells. Human LIF cDNA encodes a 202 amino acid precursor which is processed to yield a 180 amino acid, biologically active glycoprotein with an approximate molecular weight of 38-67 kDa. In addition to its involvement in the stimulation of acute-phase protein during inflammatory processes, LIF has also been implicated in promoting the survival and growth of axons in vitro. LIF signals through the LIF receptor, a heterodimer comprised of a 190 kDa LIFR subunit and the gp130 subunit common to the IL-6, IL-11, CNTF and OSM receptors. Janus tyrosine kinases, JAK1 and JAK2, are activated in response to LIF stimulation, which in turn phosphorylate Stat1 and Stat3 causing their nuclear translocation and the transcription of Stat-responsive genes.

Application

Detect Human Leukemia Inhibitory Factor using this Anti-Human Leukemia Inhibitory Factor Antibody, clone 4F7.2 validated for use in ELISA & WB.
Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation

Physical form

Format: Purified

Analysis Note

Control
Jurkat whole cell lysate

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
L0669MAB4307L1044
species reactivity

mouse, human

species reactivity

human

species reactivity

mouse

species reactivity

mouse

biological source

mouse

biological source

mouse

biological source

mouse

biological source

goat

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

clone

4F7.2, monoclonal

clone

9824, monoclonal

clone

2H2.2, monoclonal

clone

polyclonal

Gene Information

human ... LIF(3976)

Gene Information

human ... LIF(3976)

Gene Information

human ... LIF(3976)

Gene Information

mouse ... Lif(16878)

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters