Skip to Content
Merck
All Photos(1)

Documents

MAB3731

Sigma-Aldrich

Anti-Met/HGFR Antibody, phospho-specific (Tyr1234/1235), clone 6AT1877

clone 6AT1877, Chemicon®, from mouse

Synonym(s):

c-Met, cmet, HGFR, RCCP2, met proto-oncogene (hepatocyte growth factor receptor)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6AT1877, monoclonal

species reactivity

human, mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pTyr1234/pTyr1235)

Gene Information

human ... MET(4233)
mouse ... Met(17295)

Specificity

Reacts with Met/HGFR (Hepatocyte growth factor receptor) only when phosphorylated at Tyr1234 and Tyr1235. The Met receptor tyrosine kinase is the prototypical member of a small subfamily of growth factor receptors that when activated, induces mitogenic, motogenic, and morphogenic cellular responses. The ligand for Met is hepatocyte growth factor/scatter factor (HGF/SF) and while normal HGF/SF-Met signaling is required for embryonic development, abnormal Met signaling has been strongly implicated in tumorigenesis, particularly in the development of invasive and metastatic phenotypes.

Immunogen

Epitope: phospho-specific (Tyr1234/1235)
Synthetic phospho-peptide corresponding to residues surrounding Tyr1234/1235 of human Met.

Application

Anti-Met/HGFR Antibody, phospho-specific (Tyr1234/1235), clone 6AT1877 detects level of Met/HGFR & has been published & validated for use in ELISA.
ELISA (direct): 1:1,000

Optimal working dilutions must be determined by the end user.
Research Category
Signaling
Research Sub Category
Growth Factors & Receptors

Physical form

Format: Purified
Protein G purified immunoglobulin. Liquid in PBS containing 0.09% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

During shipment, small volumes of product will occasionally become entrapped in the seal of the product vial. For products with volumes of 200 μL or less, we recommend gently tapping the vial on a hard surface or briefly centrifuging the vial in a tabletop centrifuge to dislodge any liquid in the container′s cap.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service