Skip to Content
Merck

Skip To

MAB2154

Anti-P-Selectin Antibody, clone P8G6

clone P8G6, Chemicon®, from mouse

Synonym(s):

CD62P, Granule Membrane Protein-140, GMP-140, Platelet Activation Dependent Granule-external Membrane Protein, PADGEM

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μg

Estimated to ship onJune 01, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 12,800.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
P8G6, monoclonal
Application:
ELISA, ICC, IHC, IP
Citations:
3

CZK 12,800.00


Estimated to ship onJune 01, 2026Details

A recombinant, preservative-free antibody is available for your target. Try ZMS1199

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

P8G6, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable, immunocytochemistry: suitable, immunohistochemistry: suitable, immunoprecipitation (IP): suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SELP(6403)

Immunogen

IL-1beta activated HUVEC (human)

Application

Detect P-Selectin using this Anti-P-Selectin Antibody, clone P8G6 validated for use in ELISA, IP, IC, IH.
Immunohistochemistry: for use on acetone or paraformaldehyde fixed tissue

Immunocytochemistry

Immunoprecipitation

ELISA

Inhibits P Selectin dependent interactions

Inhibits platelet binding to monocytes

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)

Biochem/physiol Actions

Human P Selectin

Physical form

Format: Purified
Purified in 0.02M PBS, 0.25M NaCl, pH 7.6 with 0.1% NaN3. Liquid

Preparation Note

Maintain at 2-8°C in undiluted aliquots for up to 6 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB4700681SAB4701010SAB1406424
biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

Gene Information

human ... SELP(6403)

Gene Information

human ... SELP(6403)

Gene Information

human ... SELP(6403)

Gene Information

human ... SELP(6403)

species reactivity

human

species reactivity

nonhuman primates, human

species reactivity

-

species reactivity

human

clone

P8G6, monoclonal

clone

AK4, monoclonal

clone

HI62P, monoclonal

clone

polyclonal

antibody form

purified antibody

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

conjugate

unconjugated

conjugate

phycoerythrin (R-PE) conjugate

conjugate

unconjugated

conjugate

unconjugated


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
MAB215404053252358142

Questions

Reviews

No rating value

Active Filters