Skip to Content
Merck

Skip To

MAB1982

Anti-Integrin α6 Antibody, clone MA6

clone MA6, Chemicon®, from rat

Synonym(s):

CD49f

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg

Estimated to ship onJune 05, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 12,100.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
MA6, monoclonal
Application:
FACS, IHC, IP
Citations:
4

CZK 12,100.00


Estimated to ship onJune 05, 2026Details

A recombinant, preservative-free antibody is available for your target. Try ZRB1168

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rat

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MA6, monoclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable, immunohistochemistry: suitable, immunoprecipitation (IP): suitable

isotype

IgG2κ

suitability

not suitable for Western blot

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGA6(3655)

Application

Immunoprecipitation, flow cytometry, immunohistochemistry in frozen tissue sections. Product does not detect alpha 6 in Western blot since the epitope most likely involves both alpha 6 and beta 1 integrin subunit. Product does not block alpha 6 integrin mediated cell adhesion to laminin. Activation of integrin signaling properties by MA6 has not been characterized. Immunoprecipitation of VLA-6 from lysate of 10E6 cell equivalent requires 2 μg of MA6. Recommended concentration for flow cytometry - 10 μg/mL, staining of frozen tissue sections - 20 μg/mL.

Optimal working dilutions must be determined by end user.

Cell lines tested by using MA6 to express (6 integrins include: mouse bone marrow-derived mast cells, melanoma cell line B16F1, squamous carcinoma SCC-VII, fibroblast cell line NIH-3T3, splenic T lymphocytes, thymocytes and various T cell lines.
This Anti-Integrin α6 Antibody, clone MA6 is validated for use in FC, IP, IH for the detection of Integrin α6.

Biochem/physiol Actions

Reacts with the laminin receptor alpha-6beta-1 and alpha-6beta-4 complexes.

Physical form

Format: Purified
Liquid in 0.02M Phosphate buffer, pH 7.6, with 0.25M NaCl and 0.1% sodium azide.

Preparation Note

Store at +2-8°C for up to 12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MAB13444MABT356MAB2056
antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Gene Information

human ... ITGA6(3655)

Gene Information

human ... ITGA6(3655)

Gene Information

human ... ITGA6(3655)

Gene Information

human ... ITGA3(3675)

species reactivity

mouse

species reactivity

chicken

species reactivity

human

species reactivity

human

biological source

rat

biological source

mouse

biological source

mouse

biological source

mouse

clone

MA6, monoclonal

clone

BnC6B4, monoclonal

clone

6H7.1, monoclonal

clone

ASC-1, monoclonal


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters