Skip to Content
Merck

Skip To

IM42

Anti-MT1-MMP (Ab-3) Mouse mAb (114-6G6)

liquid, clone 114-6G6, Calbiochem®

Synonym(s):

Anti-MMP-14

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
Clone:
114-6G6, monoclonal
Species reactivity:
human
Application:
Citations:
-


biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

114-6G6, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MMP14(4323)

General description

Purified mouse monoclonal antibody generated by immunizing mice with the specified immunogen and fusing splenocytes with NS1 mouse myeloma cells. Recognizes the ~66 kDa latent and the ~60 kDa active forms of MT1-MMP protein.
Recognizes the ~60-66 kDa MTI-MMP protein in HT-1080 cells and ConA-treated MD-MB-231 cells.
This Anti-MT1-MMP (Ab-3) Mouse mAb (114-6G6) is validated for use in Frozen Sections, Paraffin Sections, Immunoblotting, Immunocytochemistry, IP for the detection of MT1-MMP (Ab-3).

Immunogen

a synthetic peptide (REVPYAYIREGHEK) corresponding to amino acids 160-173 in the catalytic domain of human MT1-MMP

Application


Frozen Sections (not recommended)
Paraffin Sections (10 g/ml, Pronase<TMSYMBOL></TMSYMBOL> or heat pre-treatment required)
Immunoblotting (10 g/ml)
Immunocytochemistry (see application reference)
Immunoprecipitation (not recommended)

Packaging

Please refer to vial label for lot-specific concentration.

Physical form

In 100 mM sodium phosphate buffer, 0.1% BSA, pH 7.0.

Preparation Note

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Negative Control
trpE (Ab-1) or untreated MD-MB-231 cells
Positive Control
HT-1080 cells, ConA-treated MD-MB-231 cells, or breast carcinoma tissue

Other Notes

Does not cross-react with MT2-MMP or MT3-MMP. To induce MT1-MMP in MD-MB-231 cells, treat with 1-5 µg/ml ConA for 24-48 h. For best results with paraffin sections, fix with perioddate-lysine-4% paraformaldehyde. Antibody should be titrated for optimal results in individual systems.
Mattei, M.G., et al. 1997. Genomics40, 168.
Okada, A., et al. 1997. J. Cell. Biol.147, 67.
Okumura, Y., et al. 1997. FEBS Lett.402, 181.
Ueno, H., et al. 1997. Cancer Res.57, 2055.
Strongin, A.Y., et al. 1995. J. Biol. Chem.270, 5331.
Takino, T., et al. 1995. J. Biol. Chem.270, 23013.
Sato, H., et al. 1994. Nature370, 61.
Cottam, D.W. and Rees, R.C. 1993. Intl J. Oncol.2, 861.
Stetler-Stevenson, W.G., et al. 1993. FASEB J.7, 1434.
Fridman, R., et al. 1992. J. Biol. Chem.267, 15389.
Woessner, J.F. 1991. FASEB J.5, 2145.
Liotta, L.A. and Stetler-Stevenson, W.G. 1990. in Sem. Cancer Biol., ed. M.M. Gottesman. Vol. 1(2):99-106.
Goldberg, G.I., et al. 1989. PNAS86, 8207.

Legal Information

Manufactured by Daiichi Fine Chemical Co., Ltd. Not available for sale in Japan.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
IM42-100UG04055977220438

Questions

Reviews

No rating value

Active Filters