Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
UNSPSC Code:
12352203
Clone:
polyclonal
Species reactivity:
human
Application:
—
Citations:
9
Pricing and availability is not currently available.
biological source
rabbit
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
polyclonal
form
liquid
contains
≤0.1% sodium azide as preservative
species reactivity
human
manufacturer/tradename
Calbiochem®
storage condition
do not freeze
isotype
IgG
shipped in
wet ice
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
human ... CTSD(1509)
General description
Anti-Cathepsin D (Ab-2), rabbit polyclonal, recognizes the ~52 kDa latent form and ~34 kDa subunit in breast carcinoma tissue. It is validated for WB and IHC with frozen and paraffin sections.
Purified rabbit polyclonal antibody. Recognizes the ~52 kDa latent form and the ~34 kDa (but not the ~14 kDa) active subunit of cathepsin D.
Recognizes the ~52 kDa latent form D and the ~34 kDa subunit, but not the ~14 kDa subunit of cathepsin D in breast carcinoma tissue extracts.
Immunogen
Human
purified human liver cathepsin D
Application
Frozen Sections (5 µg/ml)
Immunoblotting (2.5 µg/ml)
Paraffin Sections (5 µg/ml, trypsin pre-treatment required)
Immunoblotting (2.5 µg/ml)
Paraffin Sections (5 µg/ml, trypsin pre-treatment required)
Packaging
Please refer to vial label for lot-specific concentration.
Physical form
In 50 mM sodium phosphate buffer, 0.2% gelatin, pH 7.5.
Analysis Note
Positive Control
HT1080 concentrated conditioned medium or breast carcinoma tissue
HT1080 concentrated conditioned medium or breast carcinoma tissue
Other Notes
For optimal results in immunoblotting, conditioned media from HT1080 cells should be concentrated 10-20X prior to loading. Antibody should be titrated for optimal results in individual systems.
Rochefort, H. 1992. Eur. J. Cancer28A, 1780.
Rochefort, H. 1990. In Seminars in Cancer Biology, ed. M. M. Gottesman. Vol. 1(2), 153.
Westly, B. and Rochefort, H. 1980. Cell20, 352.
Barret, A. J. 1977. In Proteinases in Mammalian Cells and Tissues, ed. Barret, A. J., North Holland, New York, p 209.
Rochefort, H. 1990. In Seminars in Cancer Biology, ed. M. M. Gottesman. Vol. 1(2), 153.
Westly, B. and Rochefort, H. 1980. Cell20, 352.
Barret, A. J. 1977. In Proteinases in Mammalian Cells and Tissues, ed. Barret, A. J., North Holland, New York, p 209.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Toxicity: Standard Handling (A)
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
10-13 - German Storage Class 10 to 13
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Espen Melum et al.
Nature immunology, 20(12), 1644-1655 (2019-10-23)
Invariant natural killer T (iNKT) cells recognize activating self and microbial lipids presented by CD1d. CD1d can also bind non-activating lipids, such as sphingomyelin. We hypothesized that these serve as endogenous regulators and investigated humans and mice deficient in acid
Vaishnavi Venugopalan et al.
International journal of molecular sciences, 22(11) (2021-06-03)
Cathepsin K-mediated thyroglobulin proteolysis contributes to thyroid hormone (TH) liberation, while TH transporters like Mct8 and Mct10 ensure TH release from thyroid follicles into the blood circulation. Thus, thyroid stimulating hormone (TSH) released upon TH demand binds to TSH receptors
S N Wickramasinghe et al.
British journal of haematology, 101(2), 245-250 (1998-06-03)
Ultrastructural immunocytochemical studies were performed on sections of bone marrow from three patients with beta-thalassaemia major and two patients with haemoglobin H (HbH) disease. Some sections were reacted with either a polyclonal or a monoclonal anti-human-ubiquitin antibody and the reaction