Skip to Content
Merck

Skip To

HC19SERM1-85K

MILLIPLEX® SARS-CoV-2 Antigen Panel 1 IgM

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.47
UNSPSC Code:
12352200
To order a Milliplex® kit, please search for your analyte of interest.


species reactivity

human

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex® xMAP®)

storage temp.

2-8°C

General description

"The SARS-CoV-2 coronavirus is the pathogenic cause of the human disease COVID-19. The virus particle consists of RNA encapsulated by the nucleocapsid (N) protein which is enveloped by a membrane (M) protein. The corona-like spike proteins surrounding this inner core consist of subunits S1, S2 and the receptor binding domain which binds to the human cellular ACE-2 receptor. Upon binding, the virus enters the cell, takes over the cellular machinery, reproduces, and the viral contagion continues. Viral structural proteins offer antigenic potential for an immune response. Upon viral infection the humoral immune system responds first by producing the immunoglobulin IgM, followed later by IgG which can convey long lasting immunity. IgA, a mucosal immune response, is also found in blood.By testing for IgM, IgG, and IgA in human serum and plasma, researchers may identify those who have been exposed to the SARS-CoV-2 virus and mounted some level of immune response. Researchers may further understand the immune response over the course of infection and recovery from COVID-19."MILLIPLEX® SARS-CoV-2 Antigen Panel 1 IgM is a 4-plex kit to be used for the simultaneous analysis of human serum or plasma samples for IgM antibodies which recognize any or all of the following analytes: SARS-CoV-2 Spike S1, SARS-CoV-2 Spike S2, SARS-Cov-2 RBD, Sars-CoV-2 N

Application

The SARS-CoV-2 Antigen Panel 1 IgM is a bead-based multiplex assay based on the Luminex® xMAP technology which enables the simultaneous analysis of human samples for IgM antibodies which recognize any of the following analytes: SARS-CoV-2 Spike S1, SARS-CoV-2 Spike S2, SARS-Cov-2 RBD, Sars-CoV-2 N.This is assay yields results in Median Fluorescence Intensity (MFI).Four assay control beads are included in the base kit.This is a one day assay.This assay requires 25 μL of 1:100 diluted plasma or serum.Analytes available:SARS-CoV-2 Nucleocapsid Protein (N);SARS-CoV-2 Receptor Binding Domain (RBD);SARS-CoV-2 Spike Protein Subunit 1 (S1);SARS-CoV-2 Spike Protein Subunit 2 (S2)

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Packaging

96-well plate

Preparation Note

Recommended storage for kit components is 2 - 8°C.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HC19SERG1-85KHC19SERA1-85K03-0197-00
species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

-

manufacturer/tradename

Milliplex®

manufacturer/tradename

Milliplex®

manufacturer/tradename

Milliplex®

manufacturer/tradename

SMC®

technique(s)

multiplexing: suitable

technique(s)

multiplexing: suitable

technique(s)

multiplexing: suitable

technique(s)

single molecule counting: suitable

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

detection method

fluorometric (Luminex® xMAP®)

detection method

fluorometric (Luminex® xMAP®)

detection method

fluorometric (Luminex® xMAP®)

detection method

-


pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Sens. 1

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Uncover tips and tricks to multiplexing in this guide from the experts. Learn how you can enhance the power of your research with MILLIPLEX® multiplexing, including industry guidance for immunoassays, multiplexing tips, one-day MILLIPLEX® multiplex assays for limited lab time, and more.





Questions

  1. We are running the SARS-CoV-2 IgG, IgA, and IgM kits for the first time on the MagPix. What analysis settings should be used since the kit does not include a standard? Are there any units that should be entered?

    1 answer
    1. For Qualitative Assays, the Analysis Type is "None" in xPonent. There are no units to be entered. Median values are used for the data comparison between treatment groups.

      Helpful?

Reviews

No rating value

Active Filters