Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
NACRES:
NB.24
UNSPSC Code:
41116129
Pricing and availability is not currently available.
material
stainless steel
sterilization compatibility
autoclavable compatible
manufacturer/tradename
EZ-Fit®
input
sample type cosmetics
sample type pharmaceutical(s)
sample type beer
sample type bottled water
application(s)
bioburden testing
cosmetics
environmental
food and beverages
water monitoring
shipped in
ambient
General description
EZ-Fit® Manifold′s unique design optimizes workflow and maintenance and reduces the risk of contamination. Different filtration heads fit both reusable and disposable filtration devices. Low height design increases comfort when used in laminar flow hoods. EZ-Fit® Manifold prevents biofilm formation because every single component can be removed for easy cleaning and autoclaving.
Application
The EZ-Fit® filtration Manifold is used for universal laboratory filtrations and supports the filtration of one test sample. The manifold is suitable to be used with EZ-Fit® filtration units.
Legal Information
EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
Still not finding the right product?
Explore all of our products under EZ-Fit® Manifold
Storage Class
11 - Combustible Solids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Responsible Waste Management with EZ-Fit Filtration Unit
Bottled Water Analysis
Bottled Water Analysis
Global Trade Item Number
| SKU | GTIN |
|---|---|
| EZFITEFUN1 | 04053252958946 |