Skip to Content
Merck

Skip To

ECM552

QCM Collagen Cell Invasion Assay, 24-well (8 µm), Fluorometric

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 ea
Check Cart for Availability
CZK 18,800.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.32
eCl@ss:
32161000

CZK 18,800.00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

vertebrates

Quality Level

technique(s)

cell based assay: suitable

detection method

fluorometric

Application

The CHEMICON Cell Invasion Assay Kit is ideal for evaluation of invasive tumor cells. Each CHEMICON Cell Invasion Assay Kit contains sufficient reagents for the evaluation of 24 samples. The quantitative nature of this assay is especially useful for screening of pharmacological agents.

The CHEMICON Cell Invasion Assay Kit is intended for research use only; not for diagnostic or therapeutic applications.

Packaging

24 wells

Preparation Note

Store kit materials at 2-8°C for up to their expiration date. Do not freeze.

Other Notes

Sterile 24-well Cell Invasion Plate Assembly: (Part No. 90248) Two 24-well plates with 12 collagen-coated inserts per plate (24 inserts total/kit). Cell Detachment Solution: (Part No. 90131) One bottle - 16 mL. 4X Cell Lysis Buffer: (Part No. 90130) One bottle - 16 mL. CyQuant GR Dye1: (Part No. 90132) One vial - 75 μL Forceps: (Part No. 10203) One each.

Legal Information

GR is a trademark of Sigma-Aldrich Co. LLC

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


pictograms

CorrosionEnvironment

signalword

Danger

Storage Class

10 - Combustible liquids

wgk

WGK 2

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - ED ENV 1 - Eye Dam. 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

"Recognizing both the tremendous opportunities and the challenges facing cancer research, we are dedicated to developing and refining tools and technologies for the study of cancer. With our comprehensive portfolio, including the Upstate®, Chemicon®, and Calbiochem® brands of reagents and antibodies, researchers can count on dependable, high quality solutions for analyzing all the hallmarks of cancer."





Global Trade Item Number

SKUGTIN
ECM55204053252475375