Skip to Content
Merck

CHGE71TE3

Millipore

Millipore Express® SHC 0.5/0.2µm Cartridge 10

Code 7 EPDM, Autoclavable and SIP

Synonym(s):

Millipore Express SHC Cartridge Filter 10 in. 0.5/0.2 μm Code 7 EPDM

Sign Into View Organizational & Contract Pricing

Select a Size

3 EA
CZK 34,600.00

CZK 34,600.00


Estimated to ship onAugust 11, 2025Details



Select a Size

Change View
3 EA
CZK 34,600.00

About This Item

UNSPSC Code:
23151806
eCl@ss:
32031690

CZK 34,600.00


Estimated to ship onAugust 11, 2025Details


Product Name

Millipore Express® SHC, Cartridge, pore size 0.5/0.2 μm, cartridge nominal length 10 in. (25 cm), Code 7, EPDM rubber seal

material

EPDM rubber seal
polyethersulfone
polypropylene
polypropylene support
polysulfone

Quality Level

reg. compliance

meets FDA Indirect Food Additive requirements cited in 21 CFR 177-182 (all component materials)

sterility

non-sterile

Sterilization Compatibility

autoclavable compatible
steam-in-place compatible

product line

EMPROVE® Filter
Millipore Express®

feature

hydrophilic

manufacturer/tradename

Millipore Express®

parameter

≤28.2 mL/min air diffusion at 2.8 bar (40 psig) and 23 °C (in water)
0.07 bar max. inlet pressure (1 psi) at 135 °C (Reverse)
1.0 bar max. inlet pressure (15 psi) at 135 °C (Forward)
1.7 bar max. inlet pressure (25 psi) at 80 °C (Forward)
1700 bar max. differential pressure (25 psi) at 80 °C (Forward)
2.07 bar max. inlet pressure (30 psi) at 25 °C (Reverse)
2100 mbar max. differential pressure (30 psi) at 25 °C (Reverse)
340 mbar max. differential pressure (5 psi) at 135 °C (Forward)
69 mbar max. differential pressure (1 psi) at 135 °C (Reverse)
6900 mbar max. differential pressure (100 psi) at 25 °C (Forward)
7 bar max. inlet pressure (100 psi) at 25 °C (Forward)

technique(s)

sterile filtration: suitable

L

10 in.

W

2.7 in.

cartridge nominal length

10 in. (25 cm)

diam.

6.9 cm (2.7 in.)

filtration area

0.49 m2

impurities

≤0.25 EU/mL bacterial endotoxins (LAL test, Aqueous extraction)
<0.25 EU/mL USP bacterial endotoxins (LAL test, sample aqueous extraction)

matrix

Millipore Express® SHC

pore size

0.5/0.2 μm pore size

input

sample type liquid

bubble point

≥4000 mbar (58 psig), air with water at 23 °C

cartridge code

Code 7

General description

Millipore Express® SHC filters have an on-board polyethersulfone membrane prefilter which protects the 0.2 μm membrane from premature plugging and extends filtration capacity, so you get sterilizing-grade performance with superior throughput. The CHGE71TE3 cartridge filter features a 0.5/0.2 μm pore size, a filtration area of 0.49 m2 and a Code 7 cartridge configuration.

Millipore Express® SHC filters provide you with a single solution for a range of process fluids, minimizing validation efforts and simplifying filter inventory and stocking requirements. With Millipore Express® SHC filters, you increase productivity, shorten cycle-time and reduce cost.

Application

  • Media additives
  • Column protection
  • Concentrated protein streams
  • Cell harvest (post clarification)
  • Caustics/pH adjusters
  • Buffers
  • CIP solution

Features and Benefits

  • High capacity, high flux hydrophilic PES membranes
  • Validated sterilizing-grade performance
  • Easy wetting and integrity testing
  • Superior throughput in high fouling streams, including media and protein containing solutions
  • Increased capacity for improved process efficiency and economy
  • Broad chemical compatibility across a wide pH range (1-14) — caustic compatible

Packaging

Double Easy-Open bag

Other Notes

These products are manufactured in a facility which adheres to FDA Good Manufacturing Practices.
Directions for Use

  • Organism Retention: Microorganism
  • Mode of Action: Filtration (size exclusion)
  • Application: BioProcessing
  • Intended Use: Reduction or removal of microorganism/bioburden
  • Instructions for Use: Please refer wetting instructions of Filters with MILLIPORE EXPRESS® Membrane user guide
  • Storage Statement: Please refer Filters with MILLIPORE EXPRESS® Membrane user guide
  • Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.

Preparation Note

Sterilization Method
25 forward or 22 forward and 3 reverse SIP cycles of 30 min @ 135 °C and 0.35 bar (5) or 3 forward SIP cycles of 30 min @ 135 °C and 1 bar (15 psid); 25 autoclave cycles of 60 min @ 126 °C
This product was manufactured with a Millipore Express® membrane which meets the criteria for a "non-fiber releasing" filter as defined in 21 CFR 210.3 (b) (6).

Analysis Note

Bacterial Retention
Samples were quantitatively retentive of a minimum Brevundimonas diminuta challenge concentration of 1 x 10⁷ CFU/cm² using ASTM® F838 methodology.
TOC/Conductivity
After sterilization and a controlled water flush of 20 L, samples exhibited less than 500 ppb TOC per USP <64> and less than 1.3 µS/cm per USP <64> at 25 °C.
Will meet the USP Oxidizable Substances Test requirements after a water flush of ≥2 L

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIPORE EXPRESS is a registered trademark of Merck KGaA, Darmstadt, Germany

What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service