Skip to Content
Merck
All Photos(1)

Key Documents

CBL180-I

Sigma-Aldrich

Anti-CD62E [ELAM-1] Antibody, clone 1.2B6

clone 1.2B6, from mouse

Synonym(s):

E-selectin, CD62 antigen-like family member E, CD62E, ELAM-1, Endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, Leukocyte-endothelial cell adhesion molecule 2, P-selectin, CD62 antigen-like family member P, CD62P, GMP-140, Granu

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CZK 10,300.00

CZK 10,300.00


Availability

Available to ship onJanuary 07, 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
CZK 10,300.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CZK 10,300.00


Availability

Available to ship onJanuary 07, 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1.2B6, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
activity assay: suitable
flow cytometry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SELE(6401)

Related Categories

General description

Selectins (CD62s) are single-chain transmembrane glycoproteins that function as cell adhesion molecules (CAMs) by interacting with a range of oligosaccharide structures including sialyl-Lewis X (sLeX) via their C-type lectin domain. All three known selectins (L-, E-, and P-selectin or CD62L CD62E, CD62P, respectively) share a similar overall domain structure that includes an N-terminal C-type lectin domain (calcium-dependent lectin domain), an EGF-like domain, a variable number of short consensus repeat (SCR) or Sushi domains (2, 6, and 9 for CD62L CD62E, CD62P, respectively), a transmembrane domain, and a cytoplasmic tail. The extracellular domains show a high homology among the three selectins, the transmembrane and cytoplasmic domains however are not conserved among the three. E-selectin (CD62E, ELAM-1) and P-selectin (CD62P, GMP-140, LECAM3, PADGEM) are mainly expressed on endothelial cells at inflammatory sites, whereas L-selectin (CD62L) is exclusively expressed on leukocytes.

Specificity

This antibody is specific to human antigen E-selectin (ELAM-1, 115 kDa) and P-selectin that are expressed on activated endothelial cells. The ligand is expressed on neutrophils and may function as an early part in neutrophil binding to the endothelium. This antibody inhibits neutrophil adhesion to E-selectin and P-selectin when used as F(ab′)2 fragments. Although previously thought to recognize only human CD62E, later data (Goda, K., et al., (1999). Eur. J. Immunol. 29(5):1551-1560) showed that this antibody also recognizes human CD62P by binding to a common epitope on these members of the selectin family.

Application

Anti-CD62E [ELAM-1] Antibody, clone 1.2B6 is an antibody against CD62E [ELAM-1] for use in Immunohistochemistry, ELISA, Flow Cytometry, Western Blotting, Activity Assay.
ELISA Analysis: A representative lot detected an induction of E-selectin expression on HUVECs by T-cells isolated from human PBMCs (Yarwood, H., et al. (2000). J. Leukoc. Biol. 68(2):233-242).
ELISA Analysis: A representative lot detected a time-dependent induction of E-selectin expression by IL-1 or TNF stimulation on HUVECs, but not A549 cells (Wellicome, S.M., et al. (1990). J.Immunol. 144(7):2558-2565).
Flow Cytometry Analysis: A representative lot specifically reacted with exogenously expressed human E- and P-, but not L-, selectin on the surface of transfected cells (Goda, K., et al., (1999). Eur. J. Immunol. 29(5):1551-1560).
Flow Cytometry Analysis: A representative lot detected an induction of E-selectin expression by IL-1 or TNF stimulation on HUVECs (Wellicome, S.M., et al. (1990). J.Immunol. 144(7):2558-2565).
Western Blotting Analysis: A representative lot specifically reacted with E- and P-, but not L-, selectin IgG fusion proteins (Goda, K., et al., (1999). Eur. J. Immunol. 29(5):1551-1560).
Activity Assay: A representative lot blocked the adhesion of HL-60 cells to E- or P-selectin transfected cells (Goda, K., et al., (1999). Eur. J. Immunol. 29(5):1551-1560).
Activity Assay: A representative lot blocked sialyl-Lewis X (sLeX) binding by E- and P-selectin IgG fusion proteins (Goda, K., et al., (1999). Eur. J. Immunol. 29(5):1551-1560).
Immunohistochemistry Analysis: A representative lot detected ELAM-1 immunoreactivity in acetone-fixed frozen human skin tissue sections (Groves, R.W., et al. (1991). Br. J. Dermatol. 124(2):117-123).
Immunoprecipitation Analysis: A representative lot immunoprecipitated E-selectin and associated proteins from lysates of TNF-stimulated HUVECs (Wellicome, S.M., et al. (1990). J.Immunol. 144(7):2558-2565).

Quality

Evaluated by Immunohistochemistry in human tonsil tissue.

Immunohistochemistry Analysis: A 1:50 dilution of this antibody detected CD62E [ELAM-1] in human tonsil tissue.

Target description

66 kDa calculated.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service