Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 1 mL | Estimated to ship onMay 11, 2026fromAreál Kühne+Nagel spol. s r.o. | CZK 7,130.00 |
About This Item
biological source
goat
Quality Level
conjugate
alkaline phosphatase conjugate
antibody form
affinity purified immunoglobulin
antibody product type
secondary antibodies
clone
polyclonal
species reactivity
mouse
manufacturer/tradename
Chemicon®
technique(s)
ELISA: suitable
shipped in
wet ice
target post-translational modification
unmodified
General description
Immunogen
Application
Optimal working dilutions must be determined by the end user.
Secondary & Control Antibodies
Secondary Antibodies Adsorbed for Dual Labeling
Fragment Specific Secondary Antibodies
Biochem/physiol Actions
Physical form
Preparation Note
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) ELISA: suitable | technique(s) ELISA: suitable | technique(s) ELISA: suitable, western blot: suitable | technique(s) ELISA: suitable |
| antibody form affinity purified immunoglobulin | antibody form affinity purified immunoglobulin | antibody form affinity purified immunoglobulin | antibody form affinity purified immunoglobulin |
| conjugate alkaline phosphatase conjugate | conjugate alkaline phosphatase conjugate | conjugate alkaline phosphatase conjugate | conjugate - |
| species reactivity mouse | species reactivity human | species reactivity mouse | species reactivity mouse |
| biological source goat | biological source goat | biological source goat | biological source goat |
| clone polyclonal | clone polyclonal | clone polyclonal | clone polyclonal |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
10 - Combustible liquids
wgk
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| AP503A | 04053252675164 |



