About This Item
biological source
goat
Quality Segment
conjugate
FITC conjugate
antibody form
F(ab′)2 fragment of affinity isolated antibody
antibody product type
secondary antibodies
clone
polyclonal
species reactivity
guinea pig
manufacturer/tradename
Chemicon®
technique(s)
immunofluorescence: suitable
shipped in
wet ice
target post-translational modification
unmodified
Application
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| biological source goat | biological source goat | biological source goat | biological source goat |
| technique(s) immunofluorescence: suitable | technique(s) immunofluorescence: suitable | technique(s) immunofluorescence: suitable | technique(s) immunofluorescence: suitable |
| species reactivity guinea pig | species reactivity rabbit | species reactivity rat | species reactivity mouse |
| conjugate FITC conjugate | conjugate FITC conjugate | conjugate FITC conjugate | conjugate FITC conjugate |
| antibody form F(ab′)2 fragment of affinity isolated antibody | antibody form F(ab′)2 fragment of affinity isolated antibody | antibody form F(ab′)2 fragment of affinity isolated antibody | antibody form F(ab′)2 fragment of affinity isolated antibody |
| shipped in wet ice | shipped in wet ice | shipped in wet ice | shipped in wet ice |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3
Storage Class
11 - Combustible Solids
wgk
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| AP108F | 04053252334535 |




