Skip to Content
Merck

Skip To

AP107

Rabbit Anti-Goat IgG Antibody, Fc

Chemicon®, from rabbit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
2 mg

Estimated to ship onJune 02, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 5,080.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ELISA, IP, WB
Citations:
-

CZK 5,080.00


Estimated to ship onJune 02, 2026Details


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

clone

polyclonal

species reactivity

goat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable, immunoprecipitation (IP): suitable, western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

General description

Immunoglobulin G (IgG), is one of the most abundant proteins in human serum with normal levels between 8-17 mg/mL in adult blood. IgG is important for our defence against microorganisms and the molecules are produced by B lymphocytes as a part of our adaptive immune response. The IgG molecule has two separate functions; to bind to the pathogen that elicited the response and to recruit other cells and molecules to destroy the antigen. The variability of the IgG pool is generated by somatic recombination and the number of specificities in an individual at a given time point is estimated to be 1011 variants.

Application

Rabbit anti-Goat IgG Antibody, Fc detects level of Goat IgG & has been published & validated for use in ELISA, IP & WB.
Research Category
Secondary & Control Antibodies
Research Sub Category
Fragment Specific Secondary Antibodies

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AP181AP132AP127
species reactivity

goat

species reactivity

mouse

species reactivity

rabbit

species reactivity

mouse

technique(s)

ELISA: suitable, western blot: suitable, immunoprecipitation (IP): suitable

technique(s)

ELISA: suitable, immunoprecipitation (IP): suitable, western blot: suitable

technique(s)

ELISA: suitable, immunoprecipitation (IP): suitable, western blot: suitable

technique(s)

ELISA: suitable, immunoprecipitation (IP): suitable, western blot: suitable

antibody form

purified immunoglobulin

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody form

affinity purified immunoglobulin

antibody form

affinity purified immunoglobulin

biological source

rabbit

biological source

goat

biological source

goat

biological source

goat

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters