Skip to Content
Merck

Skip To

AM03

Anti-Bak (Ab-1) Mouse mAb (TC-100)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.41
UNSPSC Code:
51111800
Species reactivity:
(human, not mouse, not rat)
Species reactivity:
-
Application:
Citations:
-
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Level

form

liquid

species reactivity

(human, not mouse, not rat)

storage condition

do not freeze

dilution

(Immunoblotting (2.5 µg/mL, chemiluminescence)
Immunocytochemistry )

isotype

IgG2a

storage temp.

2-8°C

General description

Purified mouse monoclonal antibody generated by immunizing mice with the specified immunogen and fusing splenocytes with Sp2/0 mouse myeloma cells. Recognizes the ~30 (apparent MW) kDa Bak protein.

Immunogen

Human

Application

Immunoblotting (2.5 µg/mL, chemiluminescence)Immunocytochemistry


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
AM03-100UG04055977227970