Skip to Content
Merck

Skip To

ABT63

Anti-Ena/VASP-like protein Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Ena/VASP-like protein, Ena/vasodilator-stimulated phosphoprotein-like

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μg

Estimated to ship onJune 02, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 11,800.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, WB
Citations:
1

CZK 11,800.00


Estimated to ship onJune 02, 2026Details


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

species reactivity (predicted by homology)

rat (based on 100% sequence homology), chimpanzee (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... EVL(51466)

General description

Ena (VASP-like protein) is involved in many different cellular functions including cell migration and axon guidance. Studies have shown that Ena expression is significantly increased in breast cancer and that the level of expression is associated with tumor stage. Ena has also been linked to inner ear and hair cell development.
~46 kDa observed.

Immunogen

Epitope: EVH2 region
KLH-conjugated linear peptide corresponding to the EVH2 region of human Ena/VASP-like protein.

Application

Detect Ena/VASP-like protein using this Anti-Ena/VASP-like protein Antibody validated for use in WB & IC.
Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected Ena/VASP-like protein in NIH/3T3 cells.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton

Biochem/physiol Actions

This antibody recognizes Ena/VASP-like protein at the EVH2 region.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Ramos cell lysate
Evaluated by Western Blot in Ramos cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected Ena/VASP-like protein in 10 µg of Ramos cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HPA018849SAB4200521ABN444
Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

100

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

UniProt accession no.

Q9UI08

UniProt accession no.

Q9UI08

UniProt accession no.

P50552

UniProt accession no.

P51693


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
ABT6304053252423406

Questions

Reviews

No rating value

Active Filters