Skip to Content
Merck

Skip To

ABT10

Anti-CKAP5 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Cytoskeleton-associated protein 5, Colonic and hepatic tumor over-expressed gene protein, Ch-TOG

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC
Citations:
-


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), bovine (based on 100% sequence homology), canine (based on 100% sequence homology), rat (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CKAP5(9793)

General description

226 kDa calculated.
Three isoforms are produced by alternative splicing with molecular weights of 225, 218, and 226 kDa
Cytoskeleton-associated protein 5 (CKAP5) is a member of the TOG/XMAP215 family containing heat repeats and plays a role in kinetochore mircotubule protection from depolymerization by MCAK. CKAP5 is the homolog of the Xenopus protein, XMAP25 (also known as chTog), which plays an essential role in centrosomal assembly. CKAP5 is localized in the perinuclear cytoplasm during interphase and at the spindle poles and centrosomes during mitosis. Studies show CKAP5 interacts with TACC1, TACC3, and tubulin leading to the hypothesis that it functions in spindle microtuble organization. Hepatomas and colonic tumors have shown overexpression of CKAP5.

Immunogen

KLH-conjugated linear peptide corresponding to human CKAP5.

Application

Anti-CKAP5 Antibody is an antibody against CKAP5 for use in IC.
Immunocytochemistry Analysis: A 1:250 dilution from a previous lot detected CKAP5 in RPE1 cells.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
A431 cells
Evaluated by Immunocytochemistry in A431 cells.

Immunocytochemistry Analysis: A 1:500 dilution of this antibody detected CKAP5 in A431 cells.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HPA000278HPA008410ABE1302
antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

technique(s)

immunocytochemistry: suitable

technique(s)

immunohistochemistry: 1:50-1:200, western blot: 0.04-0.4 μg/mL

technique(s)

immunoblotting: 0.04-0.4 μg/mL, immunofluorescence: 0.25-2 μg/mL, immunohistochemistry: 1:50-1:200

technique(s)

immunocytochemistry: suitable, western blot: suitable


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
ABT1004053252633973

Questions

Reviews

No rating value

Active Filters