Skip to Content
Merck

Skip To

ABS548

Anti-Eps15 Antibody

serum, from rabbit

Synonym(s):

Epidermal growth factor receptor substrate 15, Protein Eps15, Protein AF-1p

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μL

Estimated to ship onMay 26, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 9,800.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, IHC, WB
Citations:
3

CZK 9,800.00


Estimated to ship onMay 26, 2026Details


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat, mouse

technique(s)

immunocytochemistry: suitable, immunohistochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... EPS15(2060)

General description

Eps15, also known as Epidermal growth factor receptor substrate 15, Protein AF-1p, and encoded by the gene EPS15/AF1P, is an important protein involved in cell growth regulation, cell spreading, proliferation, invagination and budding. Epidermal growth factor receptor pathway substrate 15 (Eps15) has been suggested to be involved in the endocytosis of cell surface receptors, including epidermal growth factor receptor (EGFR). Eps15 also plays a role in the assembly of clathrin-coated pits and endocytosis. Eps15 is localized in the cytoplasm and plasma membrane and in particular to clathrin coated pits upon EGFR activation. Eps15 is phosphorylated at Tyr-849 by p38alpha kinase upon stimulation with EGF during endocytic processes. Eps15 is ubiquitously expressed and a chromosomal aberration involving the Eps15 gene is associated with acute leukemia.
~125 kDa observed

Immunogen

Linear peptide corresponding to human Eps15.

Application

Anti-Eps15 Antibody is an antibody against Eps15 for use in western blotting, ICC & IHC.
Research Category
Signaling
Research Sub Category
Signaling Neuroscience
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Eps15 in 10 µg of A431, HeLa, HepG2, mouse brain, and PC12 cell lysate.
Western Blotting Analysis: A representative lot detected Eps15 in NIH/3T3 cell lysate (Fazioli, F., et al. (1993). Molecular and Cellular Biology. 13(9):5814-5828).
Western Blotting Analysis: A representative lot detected Eps15 in GST cell lysate (Umasankar, P. K., et al. (2012). Nature. 14(5):488-501).
Immunocytochemistry Analysis: A representative lot detected Eps15 in HeLa cells (Hinrichsen, L., et al. (2003). JBC. 278:45160-45170).
Immunohistochemistry Analysis: A representative lot detected Eps15 in glomerulus tissue. (Hasse, S., et al. (2006). Cell Biology. 126:219-229).

Physical form

Rabbit polyclonal serum with 0.05% sodium azide.
Unpurified

Preparation Note

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Eps15 in 10 µg of HeLa cell lysate.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HPA008451HPA061431HPA019237
Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

100

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

serum

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

UniProt accession no.

P42566

UniProt accession no.

P42566

UniProt accession no.

P42566

UniProt accession no.

Q9UBC2

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
ABS54804053252976483

Questions

Reviews

No rating value

Active Filters