Skip to Content
Merck

Skip To

ABS433

Anti-IL-32 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Interleukin-32, IL-32, Natural killer cells protein 4, Tumor necrosis factor alpha-inducing factor, NK4, TAIF

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IF, IHC, WB
Citations:
-


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunofluorescence: suitable, immunohistochemistry: suitable, western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IL32(9235)

General description

Interleukin-32 (IL-32), also known as Natural killer cells protein 4 (NK4) and Tumor necrosis factor alpha-inducing factor (TAIF), may play a role in innate and adaptive immune responses. IL-32 induces various cytokines such as TNFA/TNF-alpha, MIP-2 and IL-8, and activates typical cytokine signal pathways of NF-kappa-B and p38 MAPK. IL-32 has been shown to be induced in human peripheral lymphocyte cells after mitogen stimulation and in epithelial cells by IFN gamma, and it has been suggested to play a role in autoimmune and inflammatory diseases such as rheumatoid arthritis. IL-32 is different from other known cytokine families because it does not share sequence homology with them and is highly expressed in immune tissues, existing in at least four differentially spliced isoforms. IL-32 is selectively expressed in lymphocytes and expression is more prominent in immune cells than in non-immune cells. The expression of IL-32 increased after activation of T-cells by mitogens and after activation of NK cells by IL2/interleukin-2.
~14 kDa and ~22 kDa observed. Uncharacterized bands may appear in some lysate(s). Uniprot describes 5 isoforms between ~15 kDa and ~27 kDa

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to the C-terminus of human IL-32.

Application

Anti-IL-32 Antibody is a highly specific rabbit polyclonal antibody, that targets IL-32 & has been tested in western blotting, IHC & Immunofluorescence.
Immunohistochemistry Analysis: 10 µg/mL from a representative lot detected IL-32 in human spleen tissue.

Immunofluorescence Analysis: 20 µg/mL from a representative lot detected IL-32 in human spleen cells.
Research Category
Signaling
Research Sub Category
Developmental Signaling

Biochem/physiol Actions

This antibody recognizes two isoforms at ~14 kDa and ~22 kDa at the C-terminus of IL-32.

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human spleen tissue lysate
Evaluated by Western Blot in human spleen tissue lysate.

Western Blot Analysis: 5 µg/mL of this antibody detected IL-32 in 15 µg of human spleen tissue lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PRS3751ABN213ABF108
biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

Quality Level

100

Quality Level

-

Quality Level

100

Quality Level

100

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

shipped in

wet ice

shipped in

dry ice

shipped in

wet ice

shipped in

wet ice

Gene Information

human ... IL32(9235)

Gene Information

human ... IL32(9235)

Gene Information

human ... IL12A(3592)

Gene Information

mouse ... Il33(77125)


Still not finding the right product?

Try our Product Selector Tool to narrow your options


wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

12 - Non Combustible Liquids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
ABS43304053252910166

Questions

Reviews

No rating value

Active Filters