Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 200 μL | Estimated to ship onJune 02, 2026fromAreál Kühne+Nagel spol. s r.o. | CZK 9,780.00 |
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IF, WB
Citations:
-
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
species reactivity
human
packaging
antibody small pack of 25 μL
technique(s)
immunofluorescence: suitable, western blot: suitable
isotype
IgG
UniProt accession no.
target post-translational modification
unmodified
Gene Information
human ... VPS28(51160)
General description
Vacuolar protein sorting-associated protein 28 homolog (UniProt: Q9UK41; also known as H-Vps28, ESCRT-I complex subunit VPS28) is encoded by the VPS28 gene (Gene ID: 51160) in human. VPS28 is a component of the Endosomal Sorting Complex Required for Transport (ESCRT)-I complex that regulates the vesicular trafficking process. This complex forms the machinery driving protein sorting from endosomes to lysosomes. It is located in the cell membrane and late endosome membrane. Two isoforms of VPS28 have been described that are generated by alternative splicing. ESCRT complexes play a vital role in receptor down-regulation, lysosome biogenesis, and budding of HIV. In humans, ESCRT-I complex consists of TSG101, VPS28, and one of four potential human VPS37 homologues. The main role of ESCRT-I is to recognize ubiquitinated cargo via the UEV domain of the VPS23/TSG101 subunit. The N-terminal domain of VPS28 contains a conformationally dynamic hydrophobic binding site on its surface. The C-terminal domain of VPS28 is reported to bind the ESCRT-II complex. (Ref.: Kostelansky, MS et al. (2006). Cell 125(1); 113-126).
~51 kDa observed; 25.43 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Immunogen
A full-length recombinant ESCRT-I complex subunit VPS28 (VPS28).
Application
Anti-VPS28, Cat. No. ABS1679, is a rabbit polyclonal antibody that detects ESCRT-I complex subunit VPS28 and has been tested for use in Immunofluorescence and Western Blotting.
Research Category
Signaling
Signaling
Western Blotting Analysis: A representative lot detected recombinant VPS28 protein and VPS28 in cell extracts (Stuchell, M.D., et. al. (2004). J Biol Chem. 279(34):36059-71). Immunofluorescence Analysis: A representative lot detected VPS28 in HEK293T cells transfected with VPS28 (Silvestri, L.S., et. al. (2007). J Infect Dis. 196 Suppl 2:S264-70).
Biochem/physiol Actions
This rabbit polyclonal antibody detects ESCRT-I complex subunit VPS28 in human cells.
Physical form
Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Preparation Note
Stable for 1 year at 2-8°C from date of receipt.
Analysis Note
Evaluated by Western Blotting in GST-VPS28 recombinant protein.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected recombinant GST-VPS28 protein.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected recombinant GST-VPS28 protein.
Other Notes
Concentration: Please refer to lot specific datasheet.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Explore all of our products under Anti-VPS28
— or —
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| ABS1679 | 04054839362972 |