Skip to Content
Merck

Skip To

ABS1650

Anti-Gstp1 Antibody

serum, from rabbit

Synonym(s):

Glutathione S-transferase P, Chain 7, GST 7-7, GST class-pi, GSTP1

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
4


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat

technique(s)

immunohistochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GSTP1(2950)

General description

Glutathione S-transferase P (EC 2.5.1.18; UniProt P04906; also known as Chain 7, GST 7-7, GST class-pi) is encoded by the Gstp1 gene (Gene ID 24426) in rat species. Glutathione S-transferases constitute a family of metabolic isozymes that are involved in detoxification by catalyzing the conjugation of the reduced form of glutathione (GSH) to xenobiotic substrates. The GST family can be subdivided into the cytosolic, mitochondrial, and microsomal (a.k.a. MAPEG) proteins. Mammalian GSTs exist as either homodimers or heterodimers. The individual monomers were historically characterized by their quaternary structure and size, Yf (Mr 24.5 kDa), Yk (25 kDa), Ya1 (25.5 kDa), Ya2 (25.5 kDa), Yn (26.5 kDa), Yb1 (27 kDa), Yb2 (27 kDa) and Yc (28.5 kDa). The Y designation refers to the Y fraction (as apposed to the X and Z fractions) they were found in during purification by Sephadex G75 chromatography. Two GST superfamilies exist that comprise either cytosolic or membrane-bound proteins. Evidence suggests that cytosolic GST increases bodily resistance to carcinogens, products of oxidative stress, environmental pollutants and certain anti-cancer drugs. The cytosolic GSTs are further subdivided into eight classes. namely alpha (GSTA), mu (GSTM), pi (GSTP), sigma, theta (GSTT), kappa (GSTK), zeta (GSTZ), and omega (GSTO). GSTP exists as a homodimer composed of two Yf subunits.
~25 kDa observed (Yf monomer).

Immunogen

Purified rat liver glutathione S-transferase P.

Application

Anti-Gstp1 Antibody is an antibody against Gstp1 for use in Western Blotting, Immunohistochemistry.
Research Category
Signaling
Research Sub Category
Oxidative Stress
Western Blotting Analysis: A representative lot detected glutathione S-transferase P in rat liver and human stomach tissue homogenates (Cobbe,S.C., et al. (2003). Eur. J. Gastroenterol. Hepatol. 15(1):41-47).
Western Blotting Analysis: A representative lot detected upregulated glutathione S-transferase P (GSTP1) levels in liver homogenates from rats on diets supplemented with either one of six (EQ, CMRN, BHA, DEM, t-SO, or β-NF), but not with four other phytochemicals (OPZ, I3C, BITC, or PB) tested (Kelly, V.P., et al. (2000). Cancer Res. 60(4):957–969).
Immunohistochemistry Analysis: A representative lot detected much reduced glutathione S-transferase P (GSTP1) immunoreactivity in paraffin-embedded Barrett′s oesophagus tissues when compared to normal human oesophagus tissue sections (Cobbe,S.C., et al. (2003). Eur. J. Gastroenterol. Hepatol. 15(1):41-47).
Immunohistochemistry Analysis: A representative lot detected enhanced Glutathione S-transferase P immunoreactivity in the centrilobular part of liver from rats received dietary coumarin (CMRN) using acetone-fixed, paraffin-embedded tissue sections (Kelly, V.P., et al. (2000). Cancer Res. 60(4):957–969).

Biochem/physiol Actions

Reacts with human and rat glutathione S-transferase P dimer (Yf/Yf) and monomer (Yf).

Physical form

Rabbit polyclonal antibody in serum with 0.05% sodium azide.
Unpurified

Preparation Note

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Evaluated by Western Blotting in human lung tissue lysate.

Western Blotting Analysis: A 1:5,000 dilution of this antibody detected GSTP1 Yf monomer in 10 µg of human lung tissue lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HPA019779A7340ABS1653
conjugate

unconjugated

conjugate

unconjugated

conjugate

peroxidase conjugate

conjugate

unconjugated

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

100

antibody form

serum

antibody form

affinity isolated antibody

antibody form

IgG fraction of antiserum

antibody form

serum

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

technique(s)

immunohistochemistry: suitable, western blot: suitable

technique(s)

immunoblotting: 0.04-0.4 μg/mL, immunohistochemistry: 1:500-1:1000

technique(s)

direct ELISA: 1:10,000, western blot: 1:10,000 using lysates of Escherichia coli induced to express recombinant GST

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Faisal Bin Rashed et al.
Redox biology, 41, 101905-101905 (2021-03-01)
Tumour hypoxia negatively impacts therapy outcomes and continues to be a major unsolved clinical problem. Nitroimidazoles are hypoxia selective compounds that become entrapped in hypoxic cells by forming drug-protein adducts. They are widely used as hypoxia diagnostics and have also
Faisal Bin Rashed et al.
Redox biology, 52, 102300-102300 (2022-04-18)
Solid tumours are often poorly oxygenated, which confers resistance to standard treatment modalities. Targeting hypoxic tumours requires compounds, such as nitroimidazoles (NIs), equipped with the ability to reach and become activated within diffusion limited tumour niches. NIs become selectively entrapped



Global Trade Item Number

SKUGTIN
ABS165004055977301083

Questions

Reviews

No rating value

Active Filters