Skip to Content
Merck

Skip To

ABN306

Anti-MANF Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Mesencephalic astrocyte-derived neurotrophic factor, arginine-rich mutatedin early stage tumor, ARMET, ARP

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
-


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse, human

technique(s)

immunohistochemistry: suitable, western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MANF(7873)

General description

MANF, also known as ARMET, was initially identified as a protein containing an arginine-rich region that was highly mutated in a variety of tumors. More recently it was identified as a mesencephalic astrocyte-derived neurotrophic factor with selectivity for dopaminergic neurons, similar to glial cell line-derived neurotrophic factor (GDNF) and CDNF. In rat brain slices, MANF enhanced nigral gamma-aminobutyric acid release. Like GDNF and CDNF, MANF has selective neuroprotective activity for dopaminergic neurons suggesting that it may be indicated for the treatment of Parkinson’s disease. Expression of MANF has also been shown to be induced during ER stress, suggesting that it may play a role in protein quality control during ER stress.
~18 kDa observed. The calculated molecular weight is 21 kDa, however MANF has been shown as an ~18 kDa band in western blots. (Mizobuchi, N., et al. (2007). Cell Structure and Function. 32:41-50.). Uncharacterized bands may appear at ~25 kDa and ~100 kDa in some lysates.

Immunogen

KLH-conjugated linear peptide corresponding to human MANF.

Application

Detect the MANF protein using this Anti-MANF validated for use in WB & IHC.
Immunohistochemistry Analysis: A 1:2,000 dilution from a representative lot detected MANF in human midbrain and rat brain tissues.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Biochem/physiol Actions

This antibody does not cross-react with CDNF.

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with 0.02% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
PC12 cell lysate
Evaluated by Western Blot in PC12 cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected MANF in 15 µg/mL of PC12 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB5700091SAB3500384MABN79
biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

mouse

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

shipped in

wet ice

shipped in

wet ice

shipped in

dry ice

shipped in

wet ice

Gene Information

human ... MANF(7873)

Gene Information

human ... MANF(7873)

Gene Information

human ... ARMET(7873)

Gene Information

human ... BDNF(627)


Still not finding the right product?

Try our Product Selector Tool to narrow your options


wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

12 - Non Combustible Liquids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
ABN30604053252830976

Questions

Reviews

No rating value

Active Filters