Skip to Content
Merck

Skip To

ABE496

Anti-Lysine-specific demethylase 5C Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Lysine-specific demethylase 5C, Histone demethylase JARID1C, Jumonji/ARID domain-containing protein 1C, Protein SmcX, Protein Xe169

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, IP, WB
Citations:
-
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KDM5C(8242)

General description

Lysine-specific demethylase 5C is also called Histone demethylase JARID1C, Jumonji/ARID domain-containing protein 1C, Protein SmcX and Protein Xe169. Lysine-specific demethylase 5C is a histone demethylase that specifically targets trimethylated and dimethylated, but not monomethylated, ′Lys-4′ of histone H3, making it a regulator of the histone code. Lysine-specific demethylase 5C is involved in transcriptional repression of neuronal genes by recruiting histone deacetylases and REST at neuron-restrictive silencer elements and is associated with Mental retardation, X-linked, syndromic, Claes-Jensen type (MRXSCJ). Lysine-specific demethylase 5C is widely expressed with highest levels found in brain and skeletal muscle.
~170 kDa observed. Uniprot describes four isoforms at 176 kDa (isoform 1), 175 kDa (isoform 2), 171 kDa (isoform 3), and 155 kDa (isoform 4). Uncharacterized band(s) may be observed in some cell lysates.

Immunogen

KLH-conjugated linear peptide corresponding to a region near the near the N-terminus of human Lysine-specific demethylase 5C.

Application

Detect JARID using this rabbit polyclonal antibody, Anti-Lysine-specific demethylase 5C Antibody validated for use in western blotting, IP & IHC.
Immunoprecipitation Analysis: 10 µg of this antibody immunoprecipitated Lysine-specific demethylase 5C in 10 µg of human hippocampus tissue lysate.

Immunohistochemistry Analysis: A 1:500 dilution from a representative lot detected Lysine-specific demethylase 5C in human stomach tissue.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Biochem/physiol Actions

This antibody is expected to react with all four known isoforms based upon immunogen design.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in human hippocampus tissue lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected Lysine-specific demethylase 5C in 10 µg of human hippocampus tissue lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
ABE49604053252926075

Questions

Reviews

No rating value

Active Filters