Skip to Content
Merck

Skip To

ABE2910

Anti-DNMT3L

from rabbit

Synonym(s):

DNA (cytosine-5)-methyltransferase 3-like

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 μL

Estimated to ship onJune 05, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 4,040.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, IF, WB
Citations:
-

CZK 4,040.00


Estimated to ship onJune 05, 2026Details


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

unpurified

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

packaging

antibody small pack of 25 μL

technique(s)

immunocytochemistry: suitable, immunofluorescence: suitable, western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... Dnmt3L(54427)

General description

DNA (cytosine-5)-methyltransferase 3-like (UniProt: Q9CWR8; also known as DNMT3L) is encoded by the Dnmt3l gene (Gene ID: 54427) in murine species. DNMT3L is a catalytically inactive regulatory factor of DNA methyltransferases that can either promote or inhibit DNA methylation depending on the context.
It is shown to be essential for the function of DNMT3A and DNMT3B. It activates DNMT3A and DNMT3B by binding to their catalytic domain. It acts by accelerating the binding of DNA and S-adenosyl-L-methionine (AdoMet) to the methyltransferases and dissociates from the complex after DNA binding to the methyltransferases. DNMT3L recognizes unmethylated histone H3 lysine 4 (H3K4me0) and induces de novo DNA methylation by recruitment or activation of DNMT3. In germ cells, it is required for the methylation of imprinted loci together with DNMT3A. In male germ it is essential for the methylation of retrotransposons, which prevents their mobilization. DNMT3A also plays a key role in embryonic stem cells (ESCs) by acting both as a positive and negative regulator of DNA methylation. It promotes DNA methylation of housekeeping genes together with DNMT3A and DNMT3B. It can also serve as an inhibitor of DNA methylation at the promoter of bivalent genes. It is reported to promote differentiation of ESCs into primordial germ cells by inhibiting DNA methylation at the promoter of RHOX5, thereby activating its expression. In testicular cells it is first observed in non-dividing prospermatogonia after 12.5 dpc and its level reaches its highest at about the time of birth and then rapidly declines and is non-detectable six days postpartum when most of the prospermatognia are differentiated into dividing spermatogonial stem cells.
~50 kDa observed; 47.99 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Immunogen

Epitope: unknown
His-tagged full length (421 aa) murine recombinant DNA (cytosine-5)-methyltransferase 3-like (DNMT3L).

Application

Anti-DNMT3L, Cat. No. ABE2910, is a highly specific rabbit polyclonal antibody that targets DNA (cytosine-5)-methyltransferase 3-like and has been tested in Immunocytochemistry, Immunofluorescence, and Western Blotting.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: A representative lot detected Dnmt3l in undifferentiated J1 mouse embryonic stem cells (Nimura, K., et. al. (2006). Genes Cells. 11(10):1225-37).

Immunofluorescence Analysis: A representative lot detected Dnmt3l in mouse testes (Nimura, K., et. al. (2006). Genes Cells. 11(10):1225-37) and XY germ cells at E15.5 (Kato, Y., et. al. (2016). Nat Commun. 7:11272).

Immunocytochemistry Analysis: A representative lot detected Dnmt3l in undifferentiated J1 mouse embryonic stem cells (Nimura, K., et. al. (2006). Genes Cells. 11(10):1225-37).

Biochem/physiol Actions

This rabbit polyclonal antibody detects Dnmt3l in murine cells.

Physical form

Format: Unpurified
Rabbit polyclonal antiserum with 0.05% sodium azide.
Unpurified

Preparation Note

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Evaluated by Western Blotting in mouse embryonic stem cell lysate.

Western Blotting Analysis: A 1:2,000 dilution of this antibody detected Dnmt3l in 10 µg of mouse embryonic stem cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MABE305HPA001595HPA002694
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

antibody form

unpurified

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

biological source

rabbit

biological source

rat

biological source

rabbit

biological source

rabbit

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

species reactivity

mouse

species reactivity

human

species reactivity

human

species reactivity

human

technique(s)

immunocytochemistry: suitable, western blot: suitable, immunofluorescence: suitable

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

technique(s)

immunoblotting: 0.04-0.4 μg/mL, immunofluorescence: 0.25-2 μg/mL, immunohistochemistry: 1:500-1:1000

technique(s)

immunohistochemistry: 1:200- 1:500


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters