Skip to Content
Merck

Skip To

ABE2599

Anti-BMAL1

from guinea pig

Synonym(s):

Aryl hydrocarbon receptor nuclear translocator-like protein 1, Arnt3, Brain and muscle ARNT-like 1, HIF-1beta

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ChIP, IP
Citations:
2


biological source

guinea pig

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse, rat

packaging

antibody small pack of 25 μL

technique(s)

ChIP: suitable (ChIP-chip), ChIP: suitable (ChIP-seq), immunoprecipitation (IP): suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... ARNTL(406)
mouse ... Arntl(11865)
rat ... Arntl(29657)

General description

Aryl hydrocarbon receptor nuclear translocator-like protein 1 (UniProt: Q9WTL8; also known as Arnt3, Brain and muscle ARNT-like 1, BMAL1) is encoded by the Arnt1 (also known as Bmal1) gene (Gene ID: 11865) in murine species. BMAL1 is a transcriptional activator that forms a core component of the circadian clock. These components include ARNTL/BMAL1, CLOCK, NPAS2, ARNTL2/BMAL2, PER1, PER2, PER3, CRY1, and CRY2. CLOCK or NPAS2 and ARNTL/BMAL1 or ARNTL2/BMAL2 form the positive limb of the feedback loop. They act in the form of a heterodimer and activate the transcription of core clock genes and clock-controlled genes that are involved in key metabolic processes. PER1/2/3 and CRY1/2, act as transcriptional repressors and form the negative limb of the feedback loop. They interact with the CLOCK|NPAS2-ARNTL/BMAL1|ARNTL2/BMAL2 heterodimer inhibiting its activity. BMLA1 is highly expressed in the adult brain, skeletal muscle, and heart. It shuttles between the nucleus and the cytoplasm and this nucleocytoplasmic shuttling is essential for the nuclear accumulation of CLOCK, target gene transcription, and the degradation of the CLOCK-ARNTL/BMAL1 heterodimer. BMLA1 is reported to be phosphorylated upon dimerization with CLOCK and this phosphorylation enhances its transcriptional activity and reduces the stability of the CLOCK-ARNTL/BMAL1 heterodimer by promoting its degradation. BMAL1 null-mice are reported to have reproductive problems, reduced locomotor activity, and premature aging.
~70 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Immunogen

Epitope: unknown
KLH-conjugated linear peptide corresponding to 37 amino acids from the N-terminal half of murine Aryl hydrocarbon receptor nuclear translocator-like protein 1 (BMAL1). This sequence is conserved in isoforms 2, 4, and 5.

Application

Anti-BMAL1, Cat. No. ABE2599, is a highly specific guinea pig polyclonal antibody that targets Aryl hydrocarbon receptor nuclear translocator-like protein 1 and has been tested for use in and Chromatin Immunoprecipitation (ChIP), ChIP-seq, and Immunoprecipitation.
ChIP-seq Analysis: A representative lot detected BMAL1 in Beta-TC6 cell DNA used for chip assays (Perelis, M., et. al. (2015). Science. 350(6261):aac4250).

Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot detected BMAL1 in Beta-TC6 cell DNA used for chip assays (Perelis, M., et. al. (2015). Science. 350(6261):aac4250).
Research Category
Epigenetics & Nuclear Function

Biochem/physiol Actions

This guinea-pig polyclonal antibody detects Aryl hydrocarbon receptor nuclear translocator-like protein 1 (BMAL1). It targets an epitope within 37 amino acids from the N-terminal half.

Physical form

Affinity Purified
Format: Purified
Purified guinea pig polyclonal antibody in PBS with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Immunoprecipitation in HeLa cell nuclear extract.

Immunoprecipitation Analysis: 5 µg of this antibody detected BMAL1 in HeLa cell nuclear extract.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB2500166HPA059074ABE2598M
antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

biological source

guinea pig

biological source

goat

biological source

rabbit

biological source

guinea pig

Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

100

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

technique(s)

ChIP: suitable (ChIP-chip), immunoprecipitation (IP): suitable, ChIP: suitable (ChIP-seq)

technique(s)

indirect ELISA: suitable, western blot: suitable

technique(s)

immunoblotting: 0.04-0.4 μg/mL, immunofluorescence: 0.25-2 μg/mL, immunohistochemistry: 1:200-1:500

technique(s)

ChIP: suitable (ChIP-seq), immunocytochemistry: suitable

UniProt accession no.

Q9WTL8

UniProt accession no.

O00327

UniProt accession no.

Q8WYA1

UniProt accession no.

O08785


Still not finding the right product?

Explore all of our products under Anti-BMAL1

or

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Daniel C Levine et al.
Nature metabolism, 3(12), 1621-1632 (2021-12-15)
In mammals, circadian rhythms are entrained to the light cycle and drive daily oscillations in levels of NAD+, a cosubstrate of the class III histone deacetylase sirtuin 1 (SIRT1) that associates with clock transcription factors. Although NAD+ also participates in
Daniel C Levine et al.
Molecular cell, 78(5), 835-849 (2020-05-06)
Disrupted sleep-wake and molecular circadian rhythms are a feature of aging associated with metabolic disease and reduced levels of NAD+, yet whether changes in nucleotide metabolism control circadian behavioral and genomic rhythms remains unknown. Here, we reveal that supplementation with



Global Trade Item Number

SKUGTIN
ABE259904054839339585
ABE2599-25UL04054839464706

Questions

Reviews

No rating value

Active Filters