Skip to Content
Merck

Skip To

ABE206

Anti-MLL2 (C-Term) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Histone-lysine N-methyltransferase 2D, Lysine N-methyltransferase 2D, ALL1-related protein, Myeloid/lymphoid or mixed-lineage leukemia protein 2

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
3


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KMT2D(8085)

General description

MLL2, also known as Histone-lysine N-methyltransferase 2D, or Lysine N-methyltransferase 2D or ALL1-related protein, or Myeloid/lymphoid or mixed-lineage leukemia protein 2, and encoded by the gene KMT2D/ALR/MLL2/MLL4, is a histone methyltransferase that methylates Lys-4 of histone H3 to form H3K4me. H3K4me represents a specific tag for epigenetic transcriptional activation and for instance in estrogen receptor function where MLL2 acts as coactivator for estrogen receptor by being recruited by ESR1, thereby activating transcription. MLL2 is also a component of the ASCOM/Mll2/3 complex which plays an important role during development particularly in lymphomagenesis. MLL2 is localized to the nucleus. MLL2 is expressed in most embryonic and adult tissues. Defects in MLL2 are associated with Kabuki syndrome, a congenital mental retardation syndrome with additional features, including postnatal dwarfism, facial and bone deformities. Defects are also associated with medulloblastoma a highly malignant primary brain tumor.
~75 kDa observed

Immunogen

Epitope: C-terminus
GST-tagged recombinant protein corresponding to the C-terminus of human MLL2 (C-term).

Application

Detect MLL2 (C-Term) using this Anti-MLL2 antibody (C-Term) validated for use in western blotting & IHC.
Immunohistochemistry Analysis: A 1:500 dilution from a representative lot detected MLL2 (C-term) in human breast cancer tissue.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in human testis tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected MLL2 (C-term) in 10 µg of human testis tissue lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB4200343ABE1851ABE344
biological source

rabbit

biological source

mouse

biological source

rabbit

biological source

rabbit

antibody form

affinity isolated antibody

antibody form

purified from hybridoma cell culture

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

Quality Level

100

Quality Level

-

Quality Level

100

Quality Level

-

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

-

UniProt accession no.

O14686

UniProt accession no.

-

UniProt accession no.

Q8BRH4

UniProt accession no.

Q03164

clone

polyclonal

clone

MLL2-A, monoclonal

clone

polyclonal

clone

polyclonal


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
ABE20604053252944642

Questions

Reviews

No rating value

Active Filters