Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 25 μg | Estimated to ship onMay 29, 2026fromAreál Kühne+Nagel spol. s r.o. | CZK 4,040.00 | |
| 100 μg | Estimated to ship onMay 29, 2026fromAreál Kühne+Nagel spol. s r.o. | CZK 10,100.00 |
About This Item
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
polyclonal
species reactivity
human
packaging
antibody small pack of 25 μg
technique(s)
electron microscopy: suitable, immunocytochemistry: suitable, western blot: suitable
isotype
IgG
NCBI accession no.
UniProt accession no.
shipped in
ambient
target post-translational modification
unmodified
Gene Information
human ... CEP135(9662)
General description
Immunogen
Application
Electron Microscopy Analysis: A representative lot detected CEP135 in Electron Microscopy applications (Kleylein-Sohn, J., et. al. (2007). Dev Cell. 13(2):190-202).
Immunocytochemistry Analysis: A representative lot detected CEP135 in Immunocytochemistry applications (Kleylein-Sohn, J., et. al. (2007). Dev Cell. 13(2):190-202; Guderian, G., et. al. (2010). J Cell Sci. 123(Pt 13):2163-9; Sonnen, K.F., et. al. (2012). Biol Open. 1(10):965-76).
Biochem/physiol Actions
Physical form
Analysis Note
Western Blotting Analysis: 0.5 µg/mL of this antibody detected CEP135 in 10 µg of U2OS cell lysate.
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| antibody form purified antibody | antibody form affinity isolated antibody | antibody form purified immunoglobulin | antibody form purified immunoglobulin |
| Quality Level 100 | Quality Level 100 | Quality Level - | Quality Level 100 |
| biological source rabbit | biological source rabbit | biological source mouse | biological source mouse |
| shipped in ambient | shipped in wet ice | shipped in - | shipped in ambient |
| UniProt accession no. | UniProt accession no. - | UniProt accession no. | UniProt accession no. |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
does not flash
flash_point_c
does not flash
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| ABE1857 | 04054839233180 |
| ABE1857-25UG | 04054839453700 |



