Skip to Content
Merck
All Photos(4)

Key Documents

ABE1453

Sigma-Aldrich

Anti-phospho Brd4 (Ser492/Ser494)

from rabbit, purified by affinity chromatography

Synonym(s):

Bromodomain-containing protein 4, Protein HUNK1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
CZK 3,890.00
100 μL
CZK 10,100.00

CZK 3,890.00


Estimated to ship onMay 29, 2025Details


Request a Bulk Order

Select a Size

Change View
25 μL
CZK 3,890.00
100 μL
CZK 10,100.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CZK 3,890.00


Estimated to ship onMay 29, 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

porcine (based on 100% sequence homology), mouse (based on 100% sequence homology), rat (based on 100% sequence homology)

packaging

antibody small pack of 25 μL

technique(s)

dot blot: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

phosphorylation (pSer492)

Gene Information

human ... BRD4(23476)

General description

Bromodomain-containing protein 4 (UniProt O60885; also known as MCAP, Mitotic chromosome-associated protein, Protein HUNK1) is encoded by the BRD4 (also known as HUNK1, MCAP) gene (Gene ID 23476) in human. Bromodomain-containing protein 4 is a BET (bromodomain and extra-terminal) protein family member that functions as a chromatin reader by binding acetylated lysines in histones. It plays a key role in transmission of epigenetic memory across cell divisions and transcription regulation. It remains associated with acetylated chromatin throughout the entire cell cycle and provides epigenetic memory for post-mitotic G1 gene transcription by preserving acetylated chromatin status and maintaining high-order chromatin structure. During interphase, it plays a key role in regulating the transcription of signal-inducible genes by associating with the P-TEFb complex and recruiting it to promoters. Bromodomain-containing protein 4 is required to form the transcriptionally active P-TEFb complex where it displaces negative regulators such as HEXIM1 and 7SKsnRNA complex from P-TEFb and transforms it into an active form that can then phosphorylate the C-terminal domain of RNA polymerase II. It promotes the phosphorylation of Ser2 of the C-terminal domain of RNA polymerase II. In addition to acetylated histones, Bromodomain-containing protein 4 also recognizes and binds acetylated RELA, leading to further recruitment of the P-TEFb complex and subsequent activation of NF-kappa-B. It is also reported to act as a regulator of p53/TP53-mediated transcription and following phosphorylation by casein kinase 2K2, it is recruited to p53/TP53 specific target promoters. BRD4 knockout in mice is shown to be lethal, while BRD4 function blockage by inhibitor JQ1 (Cat. No. 500586) treatment is reported to affect the transcription of critical synaptic proteins, resulting in memory deficits and decreased seizure susceptibility in mice.

Specificity

This rabbit polyclonal antibody detects Bromodomain-conntaining protein 4 phosphorylated at Ser492/Ser494. It targets an epitope of 14 amino acids in the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 14 amino acids surrounding Ser492/Ser494 from the internal N-terminal half of human phospho Brd4 protein.

Application

Anti-phospho Brd4 (Ser492/Ser494), Cat. No. ABE1453, is a highly specific rabbit polyclonal antibody that targets Bromodomain-containing protein 4 and has been tested in Dot Blot, Immunocytochemistry, Immunohistochemistry, and Western Blotting.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: A 1:500 dilution from a representative lot detected phospho Brd4 (Ser492/Ser494) in whole cell neuronal lysate (Courtesy of Erica Korb, Ph.D., Rockefeller University).

Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected phospho Brd4 (Ser492/Ser494) in cultured cortical cells from mouse (C47blac6). (Courtesy of Erica Korb, Ph.D., Rockefeller University).

Dot Blot Analysis: A representative lot detected phospho Brd4 (Ser492/Ser494) in phospho Brd4 peptides (Courtesy of Erica Korb, Ph.D., Rockefeller University).


Quality

Evaluated by Western Blotting in humans IPS cell lysate.

Western Blotting Analysis: A 1:2,000 dilution of this antibody detected phospho Brd44 (Ser492/Ser494) in 10 µg of human IPS cell lysate.

Target description

200 kDa observed, 152.22 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xuemei Li et al.
Neurochemical research, 49(5), 1254-1267 (2024-02-21)
Fibrotic scars play important roles in tissue reconstruction and functional recovery in the late stage of nervous system injury. However, the mechanisms underlying fibrotic scar formation and regulation remain unclear. Casein kinase II (CK2) is a protein kinase that regulates
Abhiraami Kannan-Sundhari et al.
Frontiers in cell and developmental biology, 8, 576654-576654 (2020-10-06)
Hair cells (HCs) play crucial roles in perceiving sound, acceleration, and fluid motion. The tonotopic architecture of the sensory epithelium recognizes mechanical stimuli and convert them into electrical signals. The expression and regulation of the genes in the inner ear
Clara Penas et al.
Nature communications, 10(1), 3028-3028 (2019-07-12)
Cerebellar neuronal progenitors undergo a series of divisions before irreversibly exiting the cell cycle and differentiating into neurons. Dysfunction of this process underlies many neurological diseases including ataxia and the most common pediatric brain tumor, medulloblastoma. To better define the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service