Skip to Content
Merck

Skip To

ABE1306

Anti-DOM3Z Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Decapping and exoribonuclease protein, DXO, Dom-3 homolog Z, DOM3Z

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μg

Estimated to ship onMay 22, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 9,800.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
-

CZK 9,800.00


Estimated to ship onMay 22, 2026Details


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat, human

species reactivity (predicted by homology)

bovine (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DXO(1797)

General description

Decapping and exoribonuclease protein (DOM3Z), also known as DXO, Dom-3 homolog Z, and encoded by the gene symbol DOM3Z/ DOM3L/ NG6, is a ribonuclease that specifically degrades pre-mRNAs with a defective 5′ end cap. DOM3Z is part of a pre-mRNA capping quality control and has decapping, pyrophosphohydrolase and 5′-3′ exonuclease activities. DOM3Z has a decapping activity toward incomplete 5′ end cap mRNAs (such as unmethylated 5′ end-capped RNA), which in return releases GpppN and 5′ end monophosphate RNA. The 5′ end monophosphate RNA is then degraded by the 5′-3′ exoribonuclease activity, enabling this enzyme to decap and degrade incompletely capped mRNAs. Additionally, DOM3Z has been implied to have RNA 5′-pyrophosphohydrolase activity and to release pyrophosphates by hydrolyzing the 5′ end triphosphate. DOM3Z is located in the nucleus, and expressed ubiquitously.
~45 kDa observed

Immunogen

KLH-conjugated linear peptide corresponding to human DOM3Z.

Application

Anti-DOM3Z is an antibody against DOM3Z for use in Western Blotting.

Analysis Note

Evaluated by Western Blotting in human pancreas tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected DOM3Z in 10 µg of human pancreas tissue lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HPA046708WH0080153M1AB6016
antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

100

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

rabbit

biological source

rabbit

biological source

mouse

biological source

rabbit

species reactivity

mouse, rat, human

species reactivity

human

species reactivity

human

species reactivity

pig, mouse, human, rat

technique(s)

western blot: suitable

technique(s)

immunofluorescence: 0.25-2 μg/mL, immunohistochemistry: 1:200-1:500

technique(s)

indirect ELISA: suitable, western blot: 1-5 μg/mL

technique(s)

immunocytochemistry: suitable, western blot: suitable


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
ABE130604055977113594

Questions

Reviews

No rating value

Active Filters