Skip to Content
Merck

Skip To

ABE1032

Anti-ERp57 Antibody

serum, from rabbit

Synonym(s):

Protein disulfide-isomerase A3, 58 kDa glucose-regulated protein, 58 kDa microsomal protein, p58, Disulfide isomerase ER-60, Endoplasmic reticulum resident protein 57, ER protein 57, ERp57, Endoplasmic reticulum resident protein 60, ER protein 60, ERp60

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ChIP, WB
Citations:
2


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat, mouse

technique(s)

ChIP: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PDIA3(2923)

General description

Endoplasmic reticulum resident protein 57 (ERp57) is also called Protein disulfide-isomerase A3 (PDIA3), 58 kDa glucose-regulated protein (GRP58), 58 kDa microsomal protein (p58), Disulfide isomerase ER-60, and Endoplasmic reticulum resident protein 60 (ERp60). ERp57 catalyzes the rearrangement of -S-S- bonds in proteins and is associated with signal transduction, nuclear protein import and apoptosis. ERp57 is expressed in the flagellum and head regions of spermatozoa.
~57 kDa observed

Immunogen

Recombinant protein corresponding to human ERp57.

Application

Chromatin Immunoprecipitation Analysis: A representative lot from an independent laboratory lot detected ERp57 in M14 cell lysates (Chichiarelli, S., et al. (2010). Arch Biochem Biophys. 494(2):178-183; Chichiarelli, S., et al. (2007). J Cell Physiol. 210(2):343-351.).

Western Blotting Analysis: A 1:2,000 dilution from a representative lot detected ERp57 in NPA, WRO, ARO and NCI cell lysates (Prof. F. Altieri, University of Rome, La Sapienza).
Detect ERp57 using this rabbit polyclonal antibody, Anti-ERp57 Antibody validated for use in western blotting & Chromatin IP (ChIP).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Physical form

Rabbit polyclonal in buffer containing serum with 0.05% sodium azide.
Unpurified

Preparation Note

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 1:1,000 dilution of this antibody detected ERp57 in 10 µg of HeLa cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB1410582SAB1405897HPA002645
Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

100

biological source

rabbit

biological source

rabbit

biological source

mouse

biological source

rabbit

antibody form

serum

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

shipped in

wet ice

shipped in

dry ice

shipped in

dry ice

shipped in

wet ice

UniProt accession no.

P30101

UniProt accession no.

P30101

UniProt accession no.

P30101

UniProt accession no.

P30101


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
ABE103204053252927935

Questions

Reviews

No rating value

Active Filters