Skip to Content
Merck

Skip To

ABC61-M

Anti-NIPP-1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Nuclear inhibitor of protein phosphatase 1, NIPP-1, Protein phosphatase 1 regulatory inhibitor subunit 8, ARD-1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μL

Estimated to ship onMay 25, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 11,500.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, WB
Citations:
-

CZK 11,500.00


Estimated to ship onMay 25, 2026Details


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

technique(s)

immunocytochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... PPP1R8(5511)

General description

Also known as ARD1 (Activator of RNA decay), NIPP-1 (nuclear inhibitor of protein phosphatase 1) is a ubiquitously expressed multifunctional nuclear scaffold protein of 39 kDa, which plays an important role in integrating transcription, RNA processing, and cell cycle progression. NIPP-1 serves as a signaling hub in the nucleus. Additionally, NIPP-1 contains at least two binding sites for PP1 and is complexed to about one-third of the nuclear pool of PP1. Normally, the NIPP-1 - PP1 complex is inactive, but can be activated following phosphorylation of NIPP-1 with PKA, CFK2, or Src family kinases. NIPP-1 also interacts directly with nucleic acids and components of polycomb repressive complexe 2 (PRC2) and functions as a PRC2-dependent transcriptional repressor.
~42 kDa observed. Three isoforms at 39 kDa (alpha isoform), 23 kDa (beta isoform), and 13 kDa (gamma isoform) are known to exist. An uncharacterized band may be observed at ~62 kDa in some nuclear extracts.

Immunogen

GST-tagged recombinant protein corresponding to human NIPP-1.

Application

Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected NIPP-1 in Raji cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology
This NIPP-1 antibody is validated for use in WB & ICC for the detection of the NIPP-1 protein.

Biochem/physiol Actions

This antibody should recognize the alpha and beta isoforms of NIPP-1.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Raji nuclear extract
Evaluated by Western Blot Analysis in Raji nuclear extract.

Western Blot Analysis: A 1:1,000 dilution from a representative lot detected NIPP-1 in 10 µg of Raji nuclear extract.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
ABN43907-121706-852
conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Quality Level

300

Quality Level

100

Quality Level

100

Quality Level

100

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

UniProt accession no.

Q12972

UniProt accession no.

O60828

UniProt accession no.

P62140

UniProt accession no.

Q96SB3

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
ABC6104053252850752

Questions

Reviews

No rating value

Active Filters