Skip to Content
Merck

Skip To

AB5574

Anti-TrpC6 Antibody

Chemicon®, from rabbit

Synonym(s):

Transient Receptor Potential-Like Channel 6, Trp6

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
50 μL

Estimated to ship onMay 25, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 11,900.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
6

CZK 11,900.00


Estimated to ship onMay 25, 2026Details


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

rat ... Trpc6(89823)

Immunogen

Peptide corresponding to amino acids 24-38 from mouse TRPC6 (Zhu et al. 1996). Accession Q61143

Application

Anti-TrpC6 Antibody is an antibody against TrpC6 for use in IH & WB.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
Western blot:1:200-1:400 using ECL on rat brain membranes.
Immunohistochemstry on mouse brain sections.
Dilutions should be made using a carrier protein such as BSA (1-3%)
Addition of 0.1-0.5% Tween-20 to the antibody solution is recommended.
Optimal working dilutions must be determined by the end user.

Biochem/physiol Actions

Recognizes TRPC6 (Transient Receptor Potential-Like Channel 6, TRP-6). Does not cross react with any other known proteins.

SPECIES REACTIVITIES: It is expected that the antibody will also work on human due to sequence conservation (15/16). Other species have not been tested.

Physical form

Affinity purified immunoglobulin. Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, and 0.05% sodium azide as a preservative. Reconstitute with 50 μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg for 5 min).

Preparation Note

Maintain lyophilized material at -20°C for up to 12 months after date of receipt. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
CONTROL ANTIGEN: Included free of charge with the antibody is 40 μg of control antigen (lyophilized powder). The stock solution of the antigen can be made up using 100 μL of sterile deionized water. For negative control, preincubate 1 μg of peptide with 1 μg of antibody for one hour at room temperature. Optimal concentrations must be determined by the end user.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB4300572MABN748SAB5200051
biological source

rabbit

biological source

rabbit

biological source

mouse

biological source

mouse

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

-

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

UniProt accession no.

Q9Y210

UniProt accession no.

Q9Y210

UniProt accession no.

Q13507

UniProt accession no.

Q9HCX4

species reactivity

rat

species reactivity

human, mouse, rat

species reactivity

mouse

species reactivity

mouse, rat, human


Still not finding the right product?

Try our Product Selector Tool to narrow your options


hcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

11 - Combustible Solids

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters