Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 100 μg | Estimated to ship onMay 18, 2026fromAreál Kühne+Nagel spol. s r.o. | CZK 12,800.00 |
About This Item
CZK 12,800.00
Estimated to ship onMay 18, 2026Details
biological source
sheep
Quality Level
conjugate
unconjugated
antibody form
affinity purified immunoglobulin
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
rat, rabbit, human, canine
manufacturer/tradename
Chemicon®
technique(s)
immunocytochemistry: suitable, western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
dry ice
target post-translational modification
unmodified
Gene Information
human ... CYCS(54205)
Application
Apoptosis & Cancer
Metabolism
Apoptosis - Additional
Enzymes & Biochemistry
Immunocytochemistry on human and rat cells.
Optimal working dilutions must be determined by the end user.
Biochem/physiol Actions
Physical form
Preparation Note
Other Notes
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| Quality Level 100 | Quality Level 200 | Quality Level 100 | Quality Level 100 |
| biological source sheep | biological source sheep | biological source rabbit | biological source rabbit |
| antibody form affinity purified immunoglobulin | antibody form whole antiserum | antibody form affinity isolated antibody | antibody form affinity isolated antibody |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| UniProt accession no. | UniProt accession no. | UniProt accession no. | UniProt accession no. |
| shipped in dry ice | shipped in dry ice | shipped in wet ice | shipped in wet ice |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Storage Class
12 - Non Combustible Liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| AB3547 | 04053252323690 |



