Skip to Content
Merck

Skip To

AB3066

Anti-Aquaporin 2 Antibody

Chemicon®, from rabbit

Synonym(s):

AQP2, AQP-CD, MCH-CD

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ELISA, IHC, WB
Citations:
9


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable, immunohistochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... AQP2(359)

Immunogen

A 15 aa C-terminal peptide from rat AQP2. This peptide is predicted to be cytoplasmic.

Application

Detect Aquaporin 2 using this Anti-Aquaporin 2 Antibody validated for use in ELISA, IH & WB.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
Western Blotting: 1-10 μg/mL for affinity pure antibody using Chemiluminescence technique. An antibody made to the AQP2 peptide detected a major band at ~29 kDa in rat kidney cortex, and 35-47 kDa in inner and outer medulla (postranslational or glycosylation) (Nielsen et al. 1993; Deen et al. 1995). A similar pattern is seen in the human kidney (Sasaki, et al. 1994).

Immunohistochemistry: 2-10 μg/mL in paraformaldehyde fixed sections of tissues. AQP2 has been localized in apical membranes (Fushimi et al. 1993; Nielsen et al. 1993; Deen et al. 1995).

ELISA: 0.5-1.0 μg/mL using 1 μg/mL control peptide/well.

Optimal working dilutions must be determined by end user.

Biochem/physiol Actions

Aquaporin 2 (AQP-CD/WCH-CD).

SPECIES REACTIVITY: The antibody has not yet been tested for cross-reactivity to other species. The 15 aa C-terminal peptide has 100% homology with mouse and 93% homology with human AQP2.

Physical form

Affinity purified immunoglobulin.Liquid in 100 mM Tris, pH 7.5 with 0.2% BSA and 0.05% sodium azide.

Preparation Note

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A4979SAB5200110SAB4501545
conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

-

antibody form

affinity purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

Gene Information

human ... AQP2(359)

Gene Information

mouse ... Aqp5(11830)
rat ... Aqp5(25241)

Gene Information

rat ... Aqp2(25386)

Gene Information

human ... AQP1(358)

technique(s)

ELISA: suitable, western blot: suitable, immunohistochemistry: suitable

technique(s)

western blot: 1:200-1:500 using mouse salivary gland and rat lung

technique(s)

indirect immunofluorescence: suitable, western blot: suitable

technique(s)

ELISA: 1:40000, western blot: 1:500-1:1000


Still not finding the right product?

Explore all of our products under Anti-Aquaporin 2 Antibody

or

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
AB306604053252364860

Questions

Reviews

No rating value

Active Filters